Skip to Content
Merck
All Photos(1)

Key Documents

88173

Supelco

Di-n-octyl phthalate

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

DNOP, Phthalic acid di-n-octyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H38O4
CAS Number:
Molecular Weight:
390.56
Beilstein:
1915994
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

density

0.980 g/mL at 20 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-8°C

SMILES string

CCCCCCCCOC(=O)c1ccccc1C(=O)OCCCCCCCC

InChI

1S/C24H38O4/c1-3-5-7-9-11-15-19-27-23(25)21-17-13-14-18-22(21)24(26)28-20-16-12-10-8-6-4-2/h13-14,17-18H,3-12,15-16,19-20H2,1-2H3

InChI key

MQIUGAXCHLFZKX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Ion Mobility: TWCCSN2 value of 217.3 Å2 [M+Na]+

The collision cross section (CCS) measurement was provided by Waters Corporation, using the SYNAPT XS mass spectrometer.
For a description and overview of how ion mobility enables the measurement of the CCS of an ion visit ims.waters.com.
Further information on the SYNAPT XS mass spectrometer can be found on the IMS microsite and product webpage.

TWCCS measurements are expected to be within 2% of this reference value.

P/N 88173 is part of the Waters Extractables & Leachables UNIFI scientific library which can be downloaded from Waters Marketplace.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xueling Wu et al.
Journal of hazardous materials, 176(1-3), 262-268 (2009-12-05)
Two bacterial strains were isolated from activated sludge using mixtures of phthalic acid esters (PAEs) as the sole source of carbon and energy. One of the isolates was identified as Gordonia sp. strain JDC-2 and the other as Arthrobacter sp.
R H Hinton et al.
Human toxicology, 4(3), 261-271 (1985-05-01)
Treatment of rats with hypolipidaemic drugs or with the plasticizer di-2-ethyl hexyl phthalate caused significant alterations in the concentration of certain plasma proteins. Certain proteins showed dose-dependent increases, in other cases the plasma concentrations fell in treated animals. The changes
B G Lake et al.
Acta pharmacologica et toxicologica, 54(3), 167-176 (1984-03-01)
The oral administration of di-n-octyl phthalate (DNOP), mono-n-octyl phthalate (MNOP), di-(2-ethylhexyl) phthalate (DEHP) and clofibrate to young male Sprague-Dawley rats for 14 days resulted in liver enlargement. Morphological examination of liver sections from DEHP and clofibrate treated rats, but not
A B DeAngelo et al.
Toxicology, 41(3), 279-288 (1986-11-01)
Di-n-octyl phthalate (DOP) is the straight chain isomer of di(2-ethylhexyl) phthalate (DEHP) which is a widely used plasticizer and an environmental contaminant. DEHP is a strong inducer of peroxisome proliferation in rat liver. This is significant since other compounds which
Manori J Silva et al.
Toxicology, 210(2-3), 123-133 (2005-04-21)
Di-n-octyl phthalate (DnOP) is a plasticizer used in polyvinyl chloride plastics, cellulose esters, and polystyrene resins. The metabolism of DnOP results in the hydrolysis of one ester linkage to produce mono-n-octyl phthalate (MnOP), which subsequently metabolizes to form oxidative metabolites.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service