Skip to Content
Merck
All Photos(1)

Key Documents

86896

Supelco

Tetrabutylammonium tetrafluoroborate

for electrochemical analysis, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(BF4)
CAS Number:
Molecular Weight:
329.27
Beilstein:
3577508
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

Quality Level

Assay

≥99.0% (T)
≥99.0%

form

crystals

mp

155-161 °C (lit.)
159-162 °C

solubility

methanol: 0.1 g/mL, clear, colorless

SMILES string

F[B-](F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.BF4/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;2-1(3,4)5/h5-16H2,1-4H3;/q+1;-1

InChI key

NNZZSJSQYOFZAM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetrabutylammonium tetrafluoroborate (TBATFB) is a quaternary ammonium salt, mainly used as electrolyte.
Visit our Sensor Applications portal to learn more.

Application

TBATFB has been used as an electrolyte to understand the paraffin graphite powder modified with sweet potato tissue (PCPET) electrode response.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biosensor based on paraffin/graphite modified with sweet potato tissue for the determination of hydroquinone in cosmetic cream in organic phase.
Cruz Vieira I, Fatibello-Filho O.
Talanta, 52(4), 681-689 (2000)
Maria S Atanassova et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 59(7), 1655-1662 (2003-04-26)
On interacting 1,10-phenanthroline (phen) with tetrabutylammonium tetrafluoroborate (Bu4N*BF4) two novel compounds have been obtained. These are the 1:1 adduct (phen)*(Bu4N*BF4) and the complex Pb(phen)(Bu4N*BF4)3(NO3)2. The synthesized substances have been characterized by UV, IR and Raman spectroscopy as well as by
E A Glasspoole et al.
Dental materials : official publication of the Academy of Dental Materials, 17(2), 127-133 (2001-02-13)
The aim of this work was to attempt formulation of a dental composite, for investigational purposes, which will release fluoride at a rate comparable to glass ionomer materials, while retaining physical properties adequate for potential use in a number of
Grigori Guitchounts et al.
Scientific reports, 10(1), 3830-3830 (2020-03-04)
A chief goal in neuroscience is to understand how neuronal activity relates to behavior, perception, and cognition. However, monitoring neuronal activity over long periods of time is technically challenging, and limited, in part, by the invasive nature of recording tools.
Jie Ke et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(35), 14768-14772 (2009-08-27)
Electrodeposition is a widely used materials-deposition technology with a number of unique features, in particular, the efficient use of starting materials, conformal, and directed coating. The properties of the solvent medium for electrodeposition are critical to the technique's applicability. Supercritical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service