Skip to Content
Merck
All Photos(1)

Documents

81392

Supelco

Poly(vinyl chloride)

Selectophore, high molecular weight

Synonym(s):

PVC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2CHCl)n
CAS Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

form

powder

viscosity

2.0-5.0 mPa.s, 3 % in THF(25 °C)

density

1.4 g/mL at 25 °C (lit.)

SMILES string

ClC=C

InChI

1S/C2H3Cl/c1-2-3/h2H,1H2

InChI key

BZHJMEDXRYGGRV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(vinyl chloride), popular as PVC, is a polymer which is prepared from vinyl chloride monomer via polymerisation. It can never be used as such and is mostly mixed with heat stabilizers, lubricants, plasticizers, fillers, and other additives. Due to its high chemical resistance property it finds wide application in construction and lining of chemical plants.
Visit our Sensor Applications portal to learn more.

Application

It has been used to fabricate Aluminum(III) selective potentiometric sensor from PVC matrix membrane which has morin ionophore.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aluminum (III) selective potentiometric sensor based on morin in poly (vinyl chloride) matrix.
Gupta, Vinod K., Ajay K. Jain, and Gaurav Maheshwari.
Talanta, 72.4, 1469-1473 (2007)
Poly (vinyl chloride).
Allsopp, Michael W., and Giovanni Vianello.
Ullmann's Encyclopedia of Industrial Chemistry (1992)
Hitomi Sano et al.
PloS one, 7(11), e50212-e50212 (2012-12-05)
Although oxygen is essential for the wound healing process, tissue hypoxia is known to stimulate angiogenesis. To explore these inconsistent findings, we estimated the influence of the oxygen environment on wound healing with our original model. Experiment 1 (Establishment of
Christian Vogel et al.
Environmental science & technology, 47(1), 563-567 (2012-11-30)
Sewage sludge ash (SSA) is a prospective phosphorus source for the future production of recycling P-fertilizers. Due to its high heavy metals contents and the relatively low P plant-availability, SSA must be treated before agricultural utilisation. In this paper SSA
Jeffrey S Souris et al.
Molecular imaging, 12(1), 28-38 (2013-01-26)
At present, there is considerable interest in the use of in vivo fluorescence and bioluminescence imaging to track the onset and progression of pathologic processes in preclinical models of human disease. Optical quantitation of such phenomena, however, is often problematic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service