Skip to Content
Merck
All Photos(1)

Key Documents

80621

Sigma-Aldrich

Piperazine

BioUltra, anhydrous, ≥99.0% (T)

Synonym(s):

1,4-Diazacyclohexane, Diethylenediamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H10N2
CAS Number:
Molecular Weight:
86.14
Beilstein:
102555
EC Number:
MDL number:
UNSPSC Code:
12161700
eCl@ss:
39160301
PubChem Substance ID:
NACRES:
NA.25

vapor pressure

0.8 mmHg ( 20 °C)

Quality Level

product line

BioUltra

Assay

≥99.0% (T)

form

solid

expl. lim.

14 %

impurities

insoluble matter, passes filter test
≤1% water

pH

11.0-12.5 (25 °C, 0.1 M in H2O)

bp

145-146 °C (lit.)

mp

109-112 °C (lit.)

solubility

H2O: 0.1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

0.1 M in H2O

UV absorption

λ: 260 nm Amax: 0.035
λ: 280 nm Amax: 0.010

SMILES string

C1CNCCN1

InChI

1S/C4H10N2/c1-2-6-4-3-5-1/h5-6H,1-4H2

InChI key

GLUUGHFHXGJENI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Eye Dam. 1 - Flam. Sol. 1 - Repr. 2 - Resp. Sens. 1B - Skin Corr. 1B - Skin Sens. 1B

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enza Lacivita et al.
Journal of medicinal chemistry, 55(14), 6375-6380 (2012-06-29)
Here we report the design, synthesis, and 5-HT(7) receptor affinity of a set of 1-(3-biphenyl)- and 1-(2-biphenyl)piperazines. The effect on 5-HT(7) affinity of various substituents on the second (distal) phenyl ring was analyzed. Several compounds showed 5-HT(7) affinities in the
Slavka Hamulakova et al.
European journal of medicinal chemistry, 55, 23-31 (2012-07-24)
New tacrine derivatives 5a-d, 6a-d with piperazino-ethyl spacer linked with corresponding secondary amines and tacrine homodimer 8 were synthesized and tested as cholinesterase inhibitors on human acetylcholinesterase (hAChE) and human plasmatic butyrylcholinesterase (hBChE). In most cases the majority of synthesized
Mitsunori Kono et al.
Bioorganic & medicinal chemistry, 21(1), 28-41 (2012-12-12)
A series of piperazine ureas was designed, synthesized, and evaluated for their potential as novel orally available fatty acid amide hydrolase (FAAH) inhibitors that are therapeutically effective against pain. We carried out an optimization study of the lead compound 3
R A Lovell
The Veterinary clinics of North America. Small animal practice, 20(2), 453-468 (1990-03-01)
Review of all reports involving anthelmintics in dogs and cats to the IAPIC between January 1, 1986 and August 10, 1988, revealed that ivermectin (extra-label use) and piperazine accounted for over 50% of the calls assessed as toxicoses and suspected
Janie Sheridan et al.
Drug and alcohol review, 26(3), 335-343 (2007-04-25)
In this Harm Reduction Digest Sheridan, Butler, Wilkins and Russell address the emergent phenomenon of so-called 'legal party pills' which have become a significant drug issue in New Zealand and elsewhere. Although banned in a number of countries, they are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service