Skip to Content
Merck
All Photos(2)

Key Documents

76899

Supelco

Temozolomide

VETRANAL®, analytical standard

Synonym(s):

3,4-Dihydro-3-methyl-4-oxoimidazo[5,1-d]-1,2,3,5-tetrazine-8-carboxamide, 3-Methyl-4-oxo-8-imidazolo[5,1-d][1,2,3,5]tetrazinecarboxamide, 4-Methyl-5-oxo-2,3,4,6,8-pentazabicyclo[4.3.0]nona-2,7,9-triene-9-carboxamide, 8-Carbamoyl-3-methylimidazo[5,1-d]-1,2,3,5-tetrazin-4(3H)-one, NSC 362856

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N6O2
CAS Number:
Molecular Weight:
194.15
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

ign. residue

≤0.2%

loss

≤0.5% loss on drying

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

2-8°C

SMILES string

CN1N=Nc2c(ncn2C1=O)C(N)=O

InChI

1S/C6H6N6O2/c1-11-6(14)12-2-8-3(4(7)13)5(12)9-10-11/h2H,1H3,(H2,7,13)

InChI key

BPEGJWRSRHCHSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Temozolomide (TMZ) is an alkylating agent of the imidazotetrazine derivatives that shows broad-spectrum antitumor activity against murine tumors.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
TMZ may be used as a reference standard for the analysis of temozolomide in serum and brain tumor samples by micellar electrokinetic capillary chromatography (MEKC), in aqueous solutions by liquid chromatography-electrospray-tandem mass spectrometry (LC-ESI-MS/MS), in active pharmaceutical ingredients (APIs) by high performance liquid chromatography (HPLC) with MS/MS and in pharmaceutical dosage form by HPLC equipped with photodiode array (PDA) detector.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of temozolomide in serum and brain tumor with micellar electrokinetic capillary chromatography.
Andrasi M, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 879(23), 2229-2233 (2011)
Development and Validation of Simultaneous Detrmination of Anastrozole and Temozolomide in Pharmaceutical Dosage Forms.
Daphal VN, et al.
International Journal of Theoretical & Applied Sciences, 4(23), 48-55 (2012)
Stability of sample solution as a crucial point during HPLC determination of chemical purity of temozolomide drug substance
Jedynak L, et al.
Journal of Pharmaceutical and Biomedical Analysis, 83(23), 19-27 (2013)
High-performance liquid chromatographic analysis and stability of anti-tumor agent temozolomide in human plasma.
Kim H, et al.
Journal of Pharmaceutical and Biomedical Analysis, 24(3), 461-468 (2001)
Multianalyte determination of 24 cytostatics and metabolites by liquid chromatography-electrospray-tandem mass spectrometry and study of their stability and optimum storage conditions in aqueous solution.
Negreira N, et al.
Talanta, 116(23), 290-299 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service