Skip to Content
Merck
All Photos(2)

Documents

74776

Supelco

Withanolide A

analytical standard

Synonym(s):

(5α,6α,7α,22R)-6,7-Epoxy-5,20,22-trihydroxy-1-oxo-ergosta-2,24-dien-26-oic acid δ-lactone, Withaniol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H38O6
CAS Number:
Molecular Weight:
470.60
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

CC1=C(C)C(=O)O[C@H](C1)[C@](C)(O)[C@H]2CC[C@H]3[C@H]4[C@H](CC[C@]23C)[C@@]5(C)C(=O)C=CC[C@]5(O)[C@H]6O[C@@H]46

InChI

1S/C28H38O6/c1-14-13-20(33-24(30)15(14)2)27(5,31)18-9-8-16-21-17(10-12-25(16,18)3)26(4)19(29)7-6-11-28(26,32)23-22(21)34-23/h6-7,16-18,20-23,31-32H,8-13H2,1-5H3/t16-,17-,18-,20+,21-,22-,23-,25-,26-,27+,28-/m0/s1

InChI key

DXWHOKCXBGLTMQ-SFQAJKIESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Withanolide A is classified under the steroidal lactone group of bioactive compounds that are majorly found in Withania somnifera (L.).

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Withanolide A may be used as an analytical standard for the determination of withanolide A in:
  • Mentat tablet, an Indian herbal medicine by UHPLC coupled to tandem mass spectrometry (MS/MS) operating with ESI, along with multivariate analysis.
  • Withania somnifera by ultra-high performance liquid chromatography-hybrid triple quadrupole linear ion trap mass spectrometer (UHPLC-QqQ LIT-MS) equipped with electrospray ionization (ESI) source and MRM detection, along with multivariate analysis as well as HPLC combined with diode array detector (DAD).
  • Withania coagulans by LC-QqQ-ESI-MS/MS.

Biochem/physiol Actions

C28-steroidal lactone that can induce nerve development and improve nervous system function. Potential therapeutic for neurologic disorders like Alzheimer′s and Parkinson′s disease. In primary rat cortical neurons, withanolide A significantly up-regulated IDE levels, a prominent enzyme involved in degrading amyloid β protein, which may help to clear excess amyloid β protein from an Alzheimer′s-inflicted brain.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: lycium withania

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Sensitive quantification of six steroidal lactones in Withania coagulans extract by UHPLC electrospray tandem mass spectrometry.
Ali A, et al.
Steroids, 104(18), 176-181 (2015)
Quantitative determination of multi markers in five varieties of Withania somnifera using ultra-high performance liquid chromatography with hybrid triple quadrupole linear ion trap mass spectrometer combined with multivariate analysis: Application to pharmaceutical dosage forms.
Chandra P, et al.
Journal of Pharmaceutical and Biomedical Analysis, 129, 419-426 (2016)
Determination of permeability, plasma protein binding, blood partitioning, pharmacokinetics and tissue distribution of Withanolide A in rats: A neuroprotective steroidal lactone.
Singh SK, et al.
Drug Development Research (2018)
Topical delivery of Withania somnifera crude extracts in niosomes and solid lipid nanoparticles.
Chinembiri TN, et al.
Pharmacognosy magazine, 13(3), S663-S663 (2017)
Immunomodulating properties of 5,20 alpha (R)-dihydroxy-6 alpha,7 alpha-epoxy-1-oxo-(5 alpha)-whitha-2,24-dienolide and solasodine.
V Bähr et al.
Planta medica, 44(1), 32-33 (1982-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service