Skip to Content
Merck
All Photos(3)

Key Documents

71945

Sigma-Aldrich

Sodium salicylate

≥99.5% (HPLC), puriss. p.a.

Synonym(s):

2-Hydroxybenzoic acid sodium salt, Salicylic acid sodium salt, Sodium 2-hydroxybenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4COONa
CAS Number:
Molecular Weight:
160.10
Beilstein:
3732792
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.21
form:
crystals
solubility:
water: 575.7 g/L at 25 °C (77 °F)

grade

puriss. p.a.

Quality Level

Assay

≥99.5% (HPLC)

form

crystals

loss

≤0.5% loss on drying, 110 °C

color

white

useful pH range

6.85 (26.3 °C)

mp

>300 °C (lit.)

solubility

water: 575.7 g/L at 25 °C (77 °F)

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤600 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤10 mg/kg
K: ≤50 mg/kg
Mg: ≤25 mg/kg
Mn: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤25 mg/kg

SMILES string

[Na+].Oc1ccccc1C([O-])=O

InChI

1S/C7H6O3.Na/c8-6-4-2-1-3-5(6)7(9)10;/h1-4,8H,(H,9,10);/q;+1/p-1

InChI key

ABBQHOQBGMUPJH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium salicylate belongs to the class of nonacetylated salicylates.

Application

Sodium salicylate may be used as a reference standard for the quantification of the analyte in serum samples using an amperometric biosensor.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

208.9 °F - Pensky-Martens closed cup

Flash Point(C)

98.3 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosby's Pocket Dictionary of Medicine, Nursing & Health Professions - E-Book, 7 (2013)
Determination of salicylate in blood serum using an amperometric biosensor based on salicylate hydroxylase immobilized in a polypyrrole?glutaraldehyde matrix
Junior RL, et al.
Talanta, 51(3), 547-557 (2000)
Handbook of Drug Interactions: A Clinical and Forensic Guide (2003)
Kui Wang et al.
Journal of medicinal chemistry, 52(20), 6402-6412 (2009-10-16)
Viologens are showing an increasing number of scientific and technical applications in addition to their use as herbicides. However, their high toxicity poses considerable risks to human health, society, and the environment. In this context, we propose a new therapeutic
Astam K Patra et al.
Journal of hazardous materials, 201-202, 170-177 (2011-12-16)
We report a highly efficient synthetic strategy for self-assembled mesoporous γ-Al(2)O(3) materials using sodium salicylate as template. The mesoporous γ-Al(2)O(3) samples synthesized following this strategy have high surface areas (231-497 m(2)g(-1)), consist of crystalline tiny spherical nanoparticles of dimensions ca.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service