Skip to Content
Merck
All Photos(4)

Key Documents

71010BC

Millipore

Assurance® GDS Listeria monocytogenes Tq Kit

suitable for PCR testing in Food and Beverages applications

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106302
NACRES:
NA.85
Pricing and availability is not currently available.

Product Name

GDS Listeria monocytogenes Tq, Molecular-based PCR assay suitable for detection of Listeria monocytogenes

Agency

PCR, Immunomagnetic Separation (IMS)
AOAC® (PTM)
Health Canada
MicroVal

Quality Level

product line

Assurance GDS®

shelf life

6‑12 mo.

packaging

pkg of 100 tests

manufacturer/tradename

BioControl Systems 71010-100

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

BioControl GDS
for use with GDS Rotor-Gene

storage temp.

2-8°C

suitability

Listeria monocytogenes

General description

Assurance® GDS, genetic detection system, for Listeria monocytogenes Tq is an automated nucleic acid amplification system for the detection of Listeria monocytogenes in food and environmental samples. The proprietary PickPen® IMS-based sample preparation procedure quickly and effectively captures Listeria monocytogenes, separating the pathogen from inhibitory background materials ensuring valid PCR reactions for all sample types, including challenging food and environmental samples. Assurance® GDS PCR technology provides two additional levels of specificity through the use of both highly specific primers and probes to ensure accurate detection of Listeria monocytogenes.

Application

GDS, genetic detection system, for Listeria monocytogenes Tq is an automated nucleic acid amplification system for the detection of Listeria monocytogenes in foods, ingredients, and environmental samples.

Features and Benefits

  • Results in 24 hours for environmental samples and select foods.
  • Single-step enrichment reduces handling time.
  • Features innovative PickPen® immunomagnetic separation (IMS) for additional specificity and faster time to results.
  • Simplified sample preparation does not require heating blocks or preparation of reagents.
  • Proprietary Rotor-Gene® Q cycler is a rotory-based cycler that ensures uniform heating and cooling of all samples.

Components

Each 100 kit includes:
  • Listeria monocytogenes Amplification Tubes Tq (0.2 mL)
  • Listeria Concentration Reagent
  • Listeria Resuspension Buffer Tq
  • Wash Solution

Testing Method

PCR, Immunomagnetic Separation (IMS)

Legal Information

AOAC is a registered trademark of AOAC International
Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany
Assurance GDS is a registered trademark of BioControl Systems, Inc.
PICKPEN is a registered trademark of Merck KGaA, Darmstadt, Germany
Rotor-Gene is a registered trademark of Qiagen GmbH

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Assurance® GDS offers integrated solution for fast and accurate food pathogen detection in microbiology testing.

Assurance® GDS offers integrated solution for fast and accurate food pathogen detection in microbiology testing.

Assurance® GDS offers integrated solution for fast and accurate food pathogen detection in microbiology testing.

Assurance® GDS offers integrated solution for fast and accurate food pathogen detection in microbiology testing.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service