Skip to Content
Merck
All Photos(2)

Documents

61352

Supelco

4-Methyl-5-thiazoleethanol

analytical standard

Synonym(s):

5-(2-Hydroxyethyl)-4-methylthiazole, Sulfurol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9NOS
CAS Number:
Molecular Weight:
143.21
Beilstein:
114249
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

refractive index

n20/D 1.550 (lit.)
n20/D 1.550±0.001

bp

135 °C/7 mmHg (lit.)

density

1.196 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

Cc1ncsc1CCO

InChI

1S/C6H9NOS/c1-5-6(2-3-8)9-4-7-5/h4,8H,2-3H2,1H3

InChI key

BKAWJIRCKVUVED-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Niederberger et al.
Gene, 171(1), 119-122 (1996-05-24)
Amiloride (Am) inhibits growth in the fission yeast Schizosaccharomyces pombe. We show that the toxic effect of this drug is relieved by low concentrations of thiamine (Th) and that the pyrimidine moiety of the Th molecule is responsible for growth
S A Petrov et al.
Ukrainskii biokhimicheskii zhurnal (1978), 59(3), 76-79 (1987-05-01)
Thiamine, thiaminepyrophosphate and 4-methyl-5-beta-oxyethylthiazole are studied for their effect on the acetylcholinesterase activity in the brain, blood plasma and cells. The activity of acetylcholinesterase in blood cells is shown to be inhibited most of all by thiamine and thiazole. Acetylcholinesterase
Biosynthesis of thiamin thiazole: determination of the regiochemistry of the S/O acyl shift by using 1,4-dideoxy-D-xylulose-5-phosphate.
Abhishek Chatterjee et al.
Angewandte Chemie (International ed. in English), 45(21), 3507-3510 (2006-04-21)
L B Filippova et al.
Voprosy meditsinskoi khimii, 26(5), 643-646 (1980-09-01)
35S-Hemineurine penetrated across the blood-brain barrier more rapidly than 35S-thiamin within the first 30 min after intravenous administration. Within 24 hrs content of 35S-thiamin in the neurones exceeded by 34% its concentration in glia and by 200% -- in myelin
E Webb et al.
Journal of bacteriology, 179(13), 4399-4402 (1997-07-01)
Thiamine pyrophosphate (TPP) is a required cofactor in Salmonella typhimurium that is generated de novo by the condensation of 4-amino-5-hydroxymethyl pyrimidine (HMP) pyrophosphate and 4-methyl-5-(beta-hydroxyethyl)-thiazole (THZ) monophosphate. The THZ and HMP moieties are independently synthesized, and labeling studies have demonstrated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service