Skip to Content
Merck
All Photos(2)

Key Documents

59867

Supelco

Sesamin

analytical standard

Synonym(s):

Fagarol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H18O6
CAS Number:
Molecular Weight:
354.35
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

C1Oc2ccc(cc2O1)[C@H]3OC[C@H]4[C@@H]3CO[C@@H]4c5ccc6OCOc6c5

InChI

1S/C20H18O6/c1-3-15-17(25-9-23-15)5-11(1)19-13-7-22-20(14(13)8-21-19)12-2-4-16-18(6-12)26-10-24-16/h1-6,13-14,19-20H,7-10H2/t13-,14-,19+,20+/m0/s1

InChI key

PEYUIKBAABKQKQ-AFHBHXEDSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Non-competitive Δ5-desaturase inhibitor.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julie Bournival et al.
Rejuvenation research, 15(3), 322-333 (2012-04-25)
Complications of diabetes are now well-known to affect sensory, motor, and autonomic nerves. Diabetes is also thought to be involved in neurodegenerative processes characteristic of several neurodegenerative diseases. Indeed, it has been acknowledged recently that hyperglycemia-induced oxidative stress contributes to
Julie Bournival et al.
Oxidative medicine and cellular longevity, 2012, 921941-921941 (2012-08-25)
A growing body of evidence indicates that the majority of Parkinson's disease (PD) cases are associated with microglia activation with resultant elevation of various inflammatory mediators and neuroinflammation. In this study, we investigated the effects of 2 natural molecules, quercetin
Suwen Su et al.
Toxicology letters, 224(2), 257-263 (2013-11-12)
Oxidative stress caused by doxorubicin (DOX) is believed to be a major underlying molecular mechanism of DOX-induced cardiotoxicity. Sesamin (Ses), an active component extracted from sesame seeds, exhibits antioxidative and anti-inflammatory effects. In the present study, possible protective mechanisms of
Wen-xing Li et al.
Food & function, 4(3), 453-460 (2012-12-15)
This study was designed to evaluate the in vivo cardioprotective effects of food-derived sesamin in spontaneously hypertensive rats (SHR). The study was performed with 17-week-old male normotensive Wistar-Kyoto rats (WKY) and SHR which are untreated or treated with orally administered
Kaori Yasuda et al.
Drug metabolism and disposition: the biological fate of chemicals, 40(10), 1917-1926 (2012-07-04)
Sesamin and episesamin are two epimeric lignans that are found in refined sesame oil. Commercially available sesamin supplements contain both sesamin and episesamin at an approximate 1:1 ratio. Our previous study clarified the sequential metabolism of sesamin by cytochrome P450

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service