Skip to Content
Merck
All Photos(1)

Key Documents

56209

Supelco

Iron Standard for ICP

TraceCERT®, 10 g/L Fe in nitric acid (nominal concentration)

Synonym(s):

Iron ICP Standard Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

concentration

10 g/L Fe in nitric acid (nominal concentration)

technique(s)

ICP: suitable

application(s)

industrial qc
pharmaceutical

format

single component solution

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.
Certified content incl. uncertainty and expiry date are given on the certificate. Information about trace impurities are stated on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application


  • Particle Size Measurement in Microspheres: The Iron Standard for ICP was crucial in measuring and detecting bound proteins on non-porous/mesoporous silica microspheres using single-particle inductively coupled plasma mass spectrometry, a method essential for precise bioanalytical applications (Miyashita et al., 2024).

  • Iron Speciation Analysis with Portable Voltammetry: Utilizing ICP iron calibration standards, advancements in portable voltammetry have been made, allowing for more precise iron speciation analysis crucial for environmental and biochemical studies (Inaudi et al., 2023).

  • Synthesis and Biological Properties of Iron Oxide Nanoparticles: Iron standards played a role in the synthesis and characterization of iron oxide nanoparticles from honey, with ICP analysis verifying the iron content, which is vital for developing novel biomedical applications (Shahid et al., 2023).

  • Determination of Metal Concentrations in Clinical Specimens: Iron ICP-MS standards facilitated the accurate determination of aluminum concentrations in biological specimens, illustrating the versatility of iron standards in various clinical laboratory settings (San Martín et al., 2022).

  • Fast and Efficient Iron Species Determination: The application of Iron Standard for ICP in a high-resolution setup enhanced the efficiency of iron species determination using HPLC, important for quick diagnostics and research in pharmaceutical sciences (Orłowska et al., 2023).

Analysis Note

10 g/L Fe in 5% nitric acid, prepared with high purity Fe metal, HNO3 and water

Other Notes

For a complete product listing of our TraceCERT range of CRMs for ICP and AAS, technical information and example certificates please visit our inorganic TraceCERT website

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher S Lacko et al.
Journal of neural engineering, 17(1), 016057-016057 (2019-10-03)
Hydrogel scaffolds hold promise for a myriad of tissue engineering applications, but often lack tissue-mimetic architecture. Therefore, in this work, we sought to develop a new technology for the incorporation of aligned tubular architecture within hydrogel scaffolds engineered from the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service