Skip to Content
Merck
All Photos(2)

Key Documents

55823

Supelco

trans-p-Coumaric acid

analytical standard

Synonym(s):

p-Coumaric acid, trans-4-Hydroxycinnamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH=CHCO2H
CAS Number:
Molecular Weight:
164.16
Beilstein:
2207383
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

214 °C (dec.) (lit.)

application(s)

food and beverages

format

neat

SMILES string

OC(=O)\C=C\c1ccc(O)cc1

InChI

1S/C9H8O3/c10-8-4-1-7(2-5-8)3-6-9(11)12/h1-6,10H,(H,11,12)/b6-3+

InChI key

NGSWKAQJJWESNS-ZZXKWVIFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

trans-p-coumaric acid is a constituent of cutin.[1] It is also found in olive processing and wine distillery effluents.[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
trans-p-coumaric acid has been used as a standard for the identification and quantitative determination of phenolic acids in defatted rice bran using high performance liquid chromatography(HPLC).[3]

Biochem/physiol Actions

Hydroxycinnamic acid found in many fruits and vegetables.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: cinnamomum curcuma hedera humulus melissa mentha salvia sambucus zingiber

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evidence for covalently attached p-coumaric acid and ferulic acid in cutins and suberins
Riley .GR and Kolattukudy.EP
Plant Physiology, 56, 650-654 (1975)
Catalytic wet oxidation of p-coumaric acid: partial oxidation intermediates, reaction pathways and catalyst leaching
Mantzavinos D, et al.
Applied Catalysis. B, Environmental, 7, 379-396 (1996)
Toshiki Furuya et al.
Applied and environmental microbiology, 78(17), 6087-6094 (2012-06-26)
Caffeic acid is a biologically active molecule that has various beneficial properties, including antioxidant, anticancer, and anti-inflammatory activities. In this study, we explored the catalytic potential of a bacterial cytochrome P450, CYP199A2, for the biotechnological production of caffeic acid. When
Anna Vallverdú-Queralt et al.
Journal of agricultural and food chemistry, 60(38), 9667-9672 (2012-09-11)
The effect of pulsed electric fields on the polyphenol profile of tomato juices was studied. First, tomatoes were subjected to moderate-intensity pulsed electric fields (MIPEFs) and then were immediately refrigerated at 4 °C for 24 h. Treated and untreated juices
Shengbao Cai et al.
Journal of agricultural and food chemistry, 60(29), 7245-7251 (2012-07-07)
The purpose of the present work is to study the pancreatic lipase inhibitory effects of different subfractions (n-hexane, ethyl acetate (EA), n-butanol, and water) from ethanol extracts of nonfermented and fungi-fermented oats and to delineate the interactions of three primary

Protocols

HPLC Analysis of Polyphenols in Nero d'Avola Red Wine on Discovery® HS C18 (UV 280 nm)

Protocol for HPLC Analysis of Flavonoids on Ascentis® RP-Amide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service