Skip to Content
Merck
All Photos(2)

Documents

52245

Supelco

Carmoisine

analytical standard

Synonym(s):

Chromotrope FB, Azorubine, Food red 3, Acid Red 14, Disodium 4-hydroxy-3-[(4-sulfo-1-naphthalenyl)azo]-1-naphthalenesulfonate, Mordant Blue 79

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H12N2Na2O7S2
CAS Number:
Molecular Weight:
502.43
Colour Index Number:
14720
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:
E Number:
E122
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

[Na+].[Na+].Oc1c(cc(c2ccccc12)S([O-])(=O)=O)\N=N\c3ccc(c4ccccc34)S([O-])(=O)=O

InChI

1S/C20H14N2O7S2.2Na/c23-20-15-8-4-3-7-14(15)19(31(27,28)29)11-17(20)22-21-16-9-10-18(30(24,25)26)13-6-2-1-5-12(13)16;;/h1-11,23H,(H,24,25,26)(H,27,28,29);;/q;2*+1/p-2/b22-21+;;

InChI key

YSVBPNGJESBVRM-ZPZFBZIMSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Carmoisine is a synthetic azo dye, commonly added to food, beverages, medicine, and cosmetics. It is frequently used alone or in combination with Allura red (E 129) and Ponceau 4R synthetic dyes. It is reported to be used for purposes where the food needs to be heat-treated post-fermentation.

Application

Carmoisine may be used as an analytical reference standard for the quantification of the analyte in the following:
  • Food samples using spectrophotometric analysis technique.
  • Sweets and beverages using differential pulse polarography.
  • Synthetic mixtures and beverages using high performance liquid chromatography with diode array detection (HPLC-DAD).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

>437.0 °F

Flash Point(C)

> 225 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spectrophotometric determination of carmoisine after cloud point extraction using Triton X-114
Karatepe A, et al.
Turkish Journal of Chemistry, 41(2), 256-262 (2017)
Four derivative spectrophotometric methods for the simultaneous determination of carmoisine and ponceau 4R in drinks and comparison with high performance liquid chromatography
Turak F, et al.
International Journal of Analytical Chemistry, 2014 (2014)
Determination of Carmoisine, Allura red and Ponceau 4R in sweets and soft drinks by Differential Pulse Polarography
Chanlon S, et al.
J. Food Compos. Anal., 18(6), 503-515 (2005)
M C Sweatman et al.
Clinical allergy, 16(4), 331-338 (1986-07-01)
We report the case of an 8.5-year-old girl with oro-facial granulomatosis associated with clinical atopy, in whom relapse of her granulomatous disorder was shown to be related to exposure to specific food additives, viz. carmoisine, sunset yellow and monosodium glutamate.
S A Marathe et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 31(10), 739-744 (1993-10-01)
The toxicity of carmoisine and of its metabolites after interaction with media components, was studied using the single cell system of Tetrahymena pyriformis. Carmoisine was not toxic to T. pyriformis. However, when autoclaved with media components such as bactopeptone or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service