Skip to Content
Merck
All Photos(1)

Documents

52184

Supelco

2-Ethylhexyl salicylate

analytical standard

Synonym(s):

2-Ethylhexyl 2-hydroxybenzoate, Octisalate, Octyl salicylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)C6H4CO2CH2CH(C2H5)(CH2)3CH3
CAS Number:
Molecular Weight:
250.33
Beilstein:
2730664
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n/D 1.501-1.503
n20/D 1.502 (lit.)

bp

189-190 °C/21 mmHg (lit.)

density

1.014 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCC(CC)COC(=O)c1ccccc1O

InChI

1S/C15H22O3/c1-3-5-8-12(4-2)11-18-15(17)13-9-6-7-10-14(13)16/h6-7,9-10,12,16H,3-5,8,11H2,1-2H3

InChI key

FMRHJJZUHUTGKE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Ethylhexyl salicylate is used as a UV filter in sunscreens.

Application

2-Ethylhexyl salicylate may be used as a reference standard for the determination of octyl salicylate as a UV filter authorized in sunscreens by high-performance liquid chromatography.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

323.6 °F - closed cup

Flash Point(C)

162 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of the UV filters worldwide authorised in sunscreens by high-performance liquid chromatography: use of cyclodextrins as mobile phase modifier.
Chisvert A, et al.
Journal of Chromatography A, 921(2), 207-215 (2001)
A DFT Investigation of the Molecular Structure and UV Absorption Spectra of 2-Ethylhexyl 2-Hydroxybenzoate (Octisalate) and Meta-Substituted 2-Ethylhexyl 2-Hydroxybenzoate: Sunscreen Applications
Punyain W.
Applied Mechanics and Materials, 855, 15-21 (2017)
Development and Validation of RP-HPLC Method for Analysis of Four UV Filters in Sunscreen Products.
NAY, et al.
International Journal of Pharmaceutical Sciences Review and Research, 23(2), 254-258 (2013)
P Clarys et al.
Journal of dermatological science, 27(2), 77-81 (2001-09-05)
Temperature influences the stratum corneum adsorption of several topically applied compounds. This study was designed to evaluate the influence of the temperature on the stratum corneum adsorption of 3 UV filters. The UV filters were solubilized in two vehicles, an
M McVean et al.
Carcinogenesis, 18(8), 1617-1622 (1997-08-01)
Ultraviolet B (UVB, 290-320 nm) exposure results in a variety of cellular insults including induction of cyclobutane pyrimidine dimers in DNA. Accumulation of these lesions can lead to mutations in critical genes and contribute to the development of nonmelanoma skin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service