Skip to Content
Merck
All Photos(1)

Key Documents

51558

Sigma-Aldrich

HEPES buffered saline

BioUltra, for molecular biology, 2× concentrate

Synonym(s):

HEPES Saline Buffer Solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

grade

Molecular Biology
for molecular biology

Quality Level

product line

BioUltra

form

liquid

quality

2× concentrate

composition

dextrose, 2.0 g/L
HEPES, 10 g/L
KCl, 0.74 g/L
NaCl, 16 g/L
Na2HPO4.2H2O, 0.27 g/L

concentration

2 ×

impurities

DNases, none detected
RNases, none detected
phosphatases, none detected
proteases, none detected

refractive index

n20/D 1.339

pH

7.2 (25 °C)

cation traces

Al: ≤1 mg/kg
Ba: ≤1 mg/kg
Bi: ≤1 mg/kg
Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
Li: ≤1 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Mo: ≤1 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sr: ≤1 mg/kg
Zn: ≤1 mg/kg

λ

neat

UV absorption

λ: 260 nm Amax: 0.06
λ: 280 nm Amax: 0.04

SMILES string

OCCN1CCN(CC1)CCS(O)(=O)=O

InChI

1S/C8H18N2O4S/c11-7-5-9-1-3-10(4-2-9)6-8-15(12,13)14/h11H,1-8H2,(H,12,13,14)

InChI key

JKMHFZQWWAIEOD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

HEPES buffered saline is a tissue culture buffer, which is used to maintain the pH of cell culture. It is water soluble and atmosphere independent. HEPES destroys magnesium in sodium chloride solutions.

Application

HEPES buffered saline has been used:
  • for the preparation of plasmid DNA
  • as a reagent to prepare transfection master mix for DNA construct vectors
  • to develop hydrogel precursor for cell leakage assay

Components

concentration (in H2O): 10 g/l HEPES; 16 g/l NaCl; 0.74 g/l KCl; 0.27 g/l Na2HPO4.2H2O; 2.0 g/l dextrose

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Calcium phosphate transfection.
Kingston RE, Chen CA, Okayama H.
Current Protocols in Immunology, 10, 10-10 (2001)
A new approach to neural cell culture for long-term studies
Potter SM and DeMarse TB
Journal of Neuroscience Methods, 110(1-2), 17-24 (2001)
Labeling embryonic mouse central nervous system cells by in utero electroporation
Tabata H and Nakajima K
Development, Growth & Differentiation, 50(6), 507-511 (2008)
J. Sambrook et al.
Molecular Cloning: A Laboratory Manual, B11 (1989)
Attachment of microstructures to single bacteria by two-photon patterning of a protein based hydrogel
Hasselmann NF and Horn W
Biomedical Physics & Engineering Express, 4(3), 035004-035004 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service