Skip to Content
Merck
All Photos(1)

Documents

49634

Supelco

γ-Tocotrienol

analytical standard

Synonym(s):

(R)-γ-Tocotrienol, [R-(E,E)]-3,4-Dihydro-2,7,8-trimethyl-2-(4,8,12-trimethyl-3,7,11-tridecatrienyl)-2H-1-benzopyran-6-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H42O2
CAS Number:
Molecular Weight:
410.63
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

biological source

Elaeis guineensis

Quality Level

grade

analytical standard

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

very light yellow to dark yellow

application(s)

pharmaceutical (small molecule)
vitamins, nutraceuticals, and natural products

format

neat

storage temp.

−20°C

SMILES string

C\C(C)=C\CC\C(C)=C\CC\C(C)=C\CC[C@]1(C)CCc2cc(O)c(C)c(C)c2O1

InChI

1S/C28H42O2/c1-20(2)11-8-12-21(3)13-9-14-22(4)15-10-17-28(7)18-16-25-19-26(29)23(5)24(6)27(25)30-28/h11,13,15,19,29H,8-10,12,14,16-18H2,1-7H3/b21-13+,22-15+/t28-/m1/s1

InChI key

OTXNTMVVOOBZCV-WAZJVIJMSA-N

General description

γ-Tocotrienol is one of the lipid-soluble isomers of the essential micronutrient vitamin E, which shows potent antioxidant activity in lipoproteins and polyunsaturated fatty acids by means of efficient free radical scavenging.

Application

γ-Tocotrienol may be used as an analytical reference standard for the determination of the analyte in crops and biological samples by chromatographic techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Influence of growth temperature on the amounts of tocopherols, tocotrienols, and gamma-oryzanol in brown rice
Britz SJ, et al.
Journal of Agricultural and Food Chemistry, 55(18), 7559-7565 (2007)
Determination of lipophilic compounds in genetically modified rice using gas chromatography-time-of-flight mass spectrometry
Kim JK, et al.
Journal of food composition and analysis, 25(1), 31-38 (2012)
Comprehensive analysis of vitamin E constituents in human plasma by liquid chromatography- mass spectrometry
Nagy K, et al.
Analytical Chemistry, 79(18), 7087-7096 (2007)
Suzana Makpol et al.
Clinics (Sao Paulo, Brazil), 67(2), 135-143 (2012-02-24)
Human diploid fibroblasts undergo a limited number of cellular divisions in culture and progressively reach a state of irreversible growth arrest, a process termed cellular aging. The beneficial effects of vitamin E in aging have been established, but studies to
Maaike Berbée et al.
Current opinion in supportive and palliative care, 6(1), 54-59 (2012-01-10)
To give an overview of promising novel agents under development for the prevention and reduction of gastrointestinal radiation injury. Currently, several novel agents are being tested as drugs to prevent or reduce gastrointestinal radiation injury. These drugs may not only

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service