Skip to Content
Merck
All Photos(1)

Key Documents

47910

Sigma-Aldrich

Fumaric acid

≥99.0% (T)

Synonym(s):

(2E)-2-Butenedioic acid, trans-Butenedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH=CHCOOH
CAS Number:
Molecular Weight:
116.07
Beilstein:
605763
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39021709
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

vapor pressure

1.7 mmHg ( 165 °C)

Quality Level

grade

purum

Assay

≥99.0% (T)

form

powder

autoignition temp.

1364 °F

expl. lim.

40 %

mp

298-300 °C (subl.) (lit.)

solubility

95% ethanol: soluble 0.46 g/10 mL, clear, colorless

functional group

carboxylic acid

SMILES string

OC(=O)\C=C\C(O)=O

InChI

1S/C4H4O4/c5-3(6)1-2-4(7)8/h1-2H,(H,5,6)(H,7,8)/b2-1+

InChI key

VZCYOOQTPOCHFL-OWOJBTEDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fumaric acid can be prepared by fermentation by employing Rhizopus species.[1][2] Recently, industrial-scale synthesis of fumaric acid from renewable feedstocks[3] and lignocellulosic biomass[4] has been proposed

Application

Fumaric acid has been used as a standard for the quantitative determination of phenolic compounds in nettle samples by HPLC.[5]
Fumaric acid may be used in the preparation of L-Lysine-fumaric acid crystals.[6] It may also be employed for the industrial manufacture of synthetic resins and eco-friendly/biodegradable polymers.[2]

Analysis Note

sublimes at 165°C at 1.7 mm pressure (Lit.)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

523.4 °F

Flash Point(C)

273 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transport and metabolism of fumaric acid in Saccharomyces cerevisiae in aerobic glucose-limited chemostat culture.
Shah, Mihir V., et al.
Yeast (2016)
High production of fumaric acid from xylose by newly selected strain Rhizopus arrhizus RH 7-13-9. Bioresource technology
Liu H, et al.
Bioresource Technology, 186, 348-350 (2015)
Vibrational spectra and second harmonic generation in molecular complexes of L-lysine with L-tartaric, D, L-malic, acetic, arsenous, and fumaric acids.
Marchewka MK, et al.
Crystal Growth & Design, 3(4), 587-592 (2003)
N Cao et al.
Applied and environmental microbiology, 62(8), 2926-2931 (1996-08-01)
An integrated system of simultaneous fermentation-adsorption for the production and recovery of fumaric acid from glucose by Rhizopus oryzae was investigated. The system was constructed such that growing Rhizopus mycelia were self-immobilized on the plastic discs of a rotary biofilm
Tahar Ghnaya et al.
Chemosphere, 90(4), 1449-1454 (2012-10-03)
The implication of organic acids in Pb translocation was studied in two species varying in shoot lead accumulation, Sesuvium portulacastrum and Brassica juncea. Citric, fumaric, malic and α-cetoglutaric acids were separated and determined by HPLC technique in shoots, roots and

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service