Skip to Content
Merck
All Photos(1)

Key Documents

46706

Supelco

Pyrimethamine

VETRANAL®, analytical standard

Synonym(s):

5-(4-Chlorophenyl)-6-ethyl-2,4-pyrimidinediamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13ClN4
CAS Number:
Molecular Weight:
248.71
Beilstein:
219864
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

CCC1=NC(N)=NC(N)=C1C2=CC=C(Cl)C=C2

InChI

1S/C12H13ClN4/c1-2-9-10(11(14)17-12(15)16-9)7-3-5-8(13)6-4-7/h3-6H,2H2,1H3,(H4,14,15,16,17)

InChI key

WKSAUQYGYAYLPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sophie Rogée et al.
Antiviral research, 161, 1-9 (2018-10-28)
Rabies virus transmits from animals to humans and causes encephalitis. Every year more than 15 million people receive a post exposure prophylaxis (PEP) treatment that is highly effective in the prevention of rabies disease. However, when clinical symptoms appear, for
Shuxian Liu et al.
Parasites & vectors, 13(1), 242-242 (2020-05-13)
Toxoplasma gondii is a zoonotic pathogen that causes toxoplasmosis and leads to serious public health problems in developing countries. However, current clinical therapeutic drugs have some disadvantages, such as serious side effects, a long course of treatment and the emergence
Xiyuan Yao et al.
Scientific reports, 10(1), 15201-15201 (2020-09-18)
Cytotoxic drugs that are mechanistically distinct from current chemotherapies are attractive components of personalized combination regimens for combatting aggressive forms of cancer. To gain insight into the cellular mechanism of a potent platinum-acridine anticancer agent (compound 1), a correlation analysis
Inbarani Naidoo et al.
Trends in parasitology, 29(10), 505-515 (2013-09-14)
Sulfadoxine-pyrimethamine (SP) is used throughout Africa for intermittent preventive treatment (IPT) of malaria, but resistance threatens its efficacy. We found marked regional differences in the genotypes responsible for SP resistance when mapping recent surveys of dihydrofolate reductase (dhfr) and dihydropteroate
Julie Gutman et al.
The Journal of infectious diseases, 208(6), 907-916 (2013-06-27)
Malaria during pregnancy is associated with low birth weight and increased perinatal mortality, especially among primigravidae. Despite increasing prevalence of malarial parasite resistance to sulfadoxine-pyrimethamine (SP), SP continues to be recommended for intermittent preventive treatment in pregnancy (IPTp). Women without

Articles

Cancer research innovations address the complexity of the disease, providing advanced technologies for varied applications.

Cancer research innovations address the complexity of the disease, providing advanced technologies for varied applications.

Biofiles reviews innovative technologies for cancer research, reflecting the complexity of the disease.

Cancer research innovations address the complexity of the disease, providing advanced technologies for varied applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service