Skip to Content
Merck
All Photos(1)

Key Documents

46468

Supelco

Monensin sodium salt hydrate

VETRANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H61NaO11 · xH2O
CAS Number:
Molecular Weight:
692.85 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
41116107
E Number:
E714
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
clinical
cosmetics
food and beverages
forensics and toxicology
personal care
pharmaceutical (small molecule)

format

neat

Mode of action

cell membrane | interferes

storage temp.

2-8°C

SMILES string

O.[Na+].CC[C@]1(CC[C@@H](O1)[C@]2(C)CC[C@]3(C[C@H](O)[C@@H](C)[C@H](O3)[C@@H](C)[C@@H](OC)[C@H](C)C([O-])=O)O2)[C@@H]4O[C@@H](C[C@@H]4C)[C@H]5OC(O)(CO)[C@H](C)C[C@@H]5C

InChI

1S/C36H62O11.Na/c1-10-34(31-20(3)16-26(43-31)28-19(2)15-21(4)36(41,18-37)46-28)12-11-27(44-34)33(8)13-14-35(47-33)17-25(38)22(5)30(45-35)23(6)29(42-9)24(7)32(39)40;/h19-31,37-38,41H,10-18H2,1-9H3,(H,39,40);/q;+1/p-1/t19-,20-,21+,22+,23-,24-,25-,26+,27+,28-,29+,30-,31+,33-,34-,35+,36-;/m0./s1

InChI key

XOIQMTLWECTKJL-FBZUZRIGSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: polyether

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - STOT RE 2

Target Organs

Heart,muscle

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sharon Cohen et al.
Molecular immunology, 54(2), 115-121 (2012-12-27)
It has been recently demonstrated that while oligosaccharide moieties of μ heavy chains in the B-cell receptor (BCR) are of the complex type as expected, those of the pre-BCR on the surface of pre-B cells contain oligosaccharide moieties of the
T L Felix et al.
Journal of animal science, 90(11), 3905-3913 (2012-06-06)
Two experiments were conducted to evaluate the effects of monensin and dried distillers grains with solubles (DDGS) on ruminal metabolism in 8 fistulated steers. In Exp. 1, treatments were (DM basis): 1) 0 mg monensin/kg diet DM, 2) 22 mg
T F Duffield et al.
Journal of animal science, 90(12), 4583-4592 (2012-08-04)
A meta-analysis of the impact of monensin on growing and finishing beef cattle was conducted after a search of the literature. A total of 40 peer-reviewed articles and 24 additional trial reports with monensin feeding in beef cattle were selected
Katia Soto-Liebe et al.
Toxicon : official journal of the International Society on Toxinology, 60(7), 1324-1334 (2012-09-18)
Paralytic shellfish poisoning (PSP) toxins are a group of naturally occurring neurotoxic alkaloids produced among several genera of primarily freshwater cyanobacteria and marine dinoflagellates. Although saxitoxin (STX) and analogs are all potent Na(+) channel blockers in vertebrate cells, the functional
C James Newbold et al.
FEMS microbiology letters, 338(2), 161-167 (2012-12-06)
Concentrations of Na(+), K(+) and Ca(2+) in the growth medium were varied within limits normally found in vivo to determine how cation concentrations affect the sensitivity of ruminal bacteria to the ionophores, monensin (a Na(+)/H(+) and K(+)/H(+) exchanger) and tetronasin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service