Skip to Content
Merck
All Photos(1)

Key Documents

45488

Supelco

Fenobucarb

PESTANAL®, analytical standard

Synonym(s):

BPMC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17NO2
CAS Number:
Molecular Weight:
207.27
Beilstein:
2052332
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCC(C)c1ccccc1OC(=O)NC

InChI

1S/C12H17NO2/c1-4-9(2)10-7-5-6-8-11(10)15-12(14)13-3/h5-9H,4H2,1-3H3,(H,13,14)

InChI key

DIRFUJHNVNOBMY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>212.0 °F

Flash Point(C)

> 100 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nguyen Thanh Tam et al.
Environmental science and pollution research international, 25(14), 13226-13234 (2016-06-03)
Organophosphates (e.g. chlorpyrifos ethyl) and carbamates (e.g. fenobucarb) are commonly used to control a wide range of pests in rice fields of the Mekong Delta in Vietnam. This study assesses the combined effect of chlorpyrifos ethyl (CPF) and fenobucarb (F)
Huan Wu et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 174, 301-306 (2016-12-18)
A convenient fluorescence quenching method for determination of Quizalofop-p-ethyl(Qpe) was proposed in this paper. Eosin Y(EY) is a red dye with strong green fluorescence (λex/λem=519/540nm). The interaction between EY, Pd(II) and Qpe was investigated by fluorescence spectroscopy, resonance Rayleigh scattering(RRS)
H Kobayashi et al.
Toxicology letters, 29(2-3), 153-159 (1985-12-01)
Mice showed no toxic signs after a single injection of o-sec-butylphenyl methylcarbamate (BPMC, 10 mg/kg) or 2-isopropoxyphenyl-N-methylcarbamate (propoxur, 2 mg/kg). Each dose of BPMC or propoxur caused an increase in acetylcholine content and a decrease in acetylcholinesterase activity in the
C L Qiao et al.
Journal of medical entomology, 36(6), 666-670 (1999-12-11)
Organophosphate (OP) insecticides have been used widely to control Culex pipiens L. populations and this has led to the emergence of OP-resistance. Predominantly, resistance in Cx. pipiens is caused by over-production of nonspecific esterases, such as Est beta 1(1) and
Yuko Ito et al.
Journal of chromatography. A, 1187(1-2), 53-57 (2008-02-26)
Dual counter-current chromatography (dual CCC)-tandem mass spectrometry (MS/MS) was successfully performed with a newly designed spiral column for dual CCC. The small column capacity required for directly coupling with electrospray MS/MS was accomplished by forming a rectangular spiral groove on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service