Skip to Content
Merck
All Photos(1)

Key Documents

36232

Supelco

Density Standard 1251 kg/m3

H&D Fitzgerald Ltd. Quality

Synonym(s):

Sodium bromide solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
3587179
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

H&D Fitzgerald Ltd.

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
industrial qc
personal care
petroleum
pharmaceutical (small molecule)

format

mixture

SMILES string

[Na+].[Br-]

InChI

1S/BrH.Na/h1H;/q;+1/p-1

InChI key

JHJLBTNAGRQEKS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

This density CRM is used for the calibration of all types of laboratory density meters in the test temperature range of 15 to 25 °C.

Features and Benefits

  • Manufactured and verified in an ISO 17025 accredited laboratory, as per UKAS
  • Carries an associated uncertainty of +/- 0.010 Kg/m3
  • Provided in a sealed glass ampoule of 10 mL to assure safe delivery
  • Accompanied with a certificate of analysis (CoA) having information on the expiry date for an effective audit

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Dragana Despotovic et al.
Journal of biomolecular screening, 17(6), 796-805 (2012-03-07)
Directed evolution offers opportunities to improve promiscuous activities of hydrolases in rounds of diversity generation and high-throughput screening. In this article, we developed and validated a screening platform to improve the perhydrolytic activity of proteases and likely other hydrolases (e.g.
Jason E DeRouchey et al.
The journal of physical chemistry. B, 115(41), 11888-11894 (2011-09-08)
Using osmotic stress coupled with X-ray scattering, we have directly examined the salt sensitivity of the intermolecular forces between helices in condensed protamine-DNA arrays. Thermodynamic forces are measured from the dependence of DNA helical interaxial spacings on external salt concentration
Catarina N Matias et al.
Biomedical chromatography : BMC, 26(11), 1348-1352 (2012-01-26)
Extracellular water (ECW) assessment is based on dilution techniques, commonly using blood sampling. However, plasma collection is an invasive procedure. We aimed to validate the use of saliva for ECW estimation by the bromide dilution technique using plasma as the
C Langdon Fielding et al.
American journal of veterinary research, 72(10), 1390-1396 (2011-10-04)
To determine values for total body water (TBW), extracellular fluid volume (ECFV), intracellular fluid volume (ICFV), and plasma volume (PV) in healthy neonatal (< 24 hours old) foals and to create a multifrequency bioelectrical impedance analysis (MF-BIA) model for use
I Iu Ponedel'kina et al.
Bioorganicheskaia khimiia, 36(3), 387-391 (2010-07-21)
The oxidation of primary hydroxyl groups in dermatan sulfate with the NaOCl-NaBr-2,2,6,6-tetramethylpiperidine-1-oxyl (TEMPO) reagent in an aqueous alkaline environment was carried out for the first time. Modified dermatan sulfates containing hydrated aldehyde (15-50%) and carboxyl (25-100%) groups were obtained.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service