Skip to Content
Merck
All Photos(1)

Documents

33973

Supelco

Clorsulon

VETRANAL®, analytical standard

Synonym(s):

4-Amino-6-(trichlorovinyl)-1,3-benzenedisulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8Cl3N3O4S2
CAS Number:
Molecular Weight:
380.66
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Nc1cc(\C(Cl)=C(\Cl)Cl)c(cc1S(N)(=O)=O)S(N)(=O)=O

InChI

1S/C8H8Cl3N3O4S2/c9-7(8(10)11)3-1-4(12)6(20(14,17)18)2-5(3)19(13,15)16/h1-2H,12H2,(H2,13,15,16)(H2,14,17,18)

InChI key

QOVTVIYTBRHADL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Clorsulon, is a benzene sulphonamide and a flukicide, which is found to be effective against the adult and immature form of liver fluke, Fasciola hepatica in cattle.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S F Sundlof et al.
Journal of veterinary pharmacology and therapeutics, 15(3), 282-291 (1992-09-01)
Clorsulon was measured in plasma and urine of sheep and goats after administration of a single intravenous (i.v.) and after a single oral dose of 7 mg/kg. A three-compartment model with elimination occurring from the central compartment was determined to
Y Vera Montenegro et al.
Parasitology research, 91(1), 1-4 (2003-07-05)
The efficacy of 5-chloro-2-methylthio-6-(1-napthyloxy)-1H-benzimidazole was evaluated with three commercial fasciolicides in terms of the percentage of egg reduction in cattle. Fifty Swiss cows were selected for inclusion in the trial based on finding eggs of Fasciola hepatica in their feces.
Induction of reversible urothelial cell hyperplasia in rats by clorsulon, a flukicide with weak carbonic anhydrase inhibitory activity
Lankas.RG and Peter.PC
Food And Chemical Toxicology, 30, 297-306 (1992)
T A Morsy et al.
Journal of the Egyptian Society of Parasitology, 31(1), 79-85 (2003-02-01)
Scabies infestation is a community health problem worldwide. This is particularly true in overcrowded and unhygienic areas. Seven dogs were experimentally infested with Sarcoptes scabiei freshly recovered from a patient. The dogs after infestation were treated with a mixture of
L Moll et al.
Veterinary parasitology, 91(1-2), 153-158 (2000-07-13)
In the winter of 1998/1999, sheep on a farm in the province of North Holland, The Netherlands, died from subacute and chronic liver fluke disease despite four previous treatments with triclabendazole (TCBZ). Faecal examinations of sheep and cattle on the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service