Skip to Content
Merck
All Photos(2)

Documents

336092

Sigma-Aldrich

1-Methylimidazole

≥99%, purified by redistillation

Synonym(s):

N-Methylimidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N2
CAS Number:
Molecular Weight:
82.10
Beilstein:
105197
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

vapor pressure

0.4 mmHg ( 20 °C)

Quality Level

Assay

≥99%

form

liquid

autoignition temp.

977 °F

purified by

redistillation

expl. lim.

15.7 %

impurities

≤19 ppm Triazine
<0.005% water

refractive index

n20/D 1.495 (lit.)

bp

198 °C (lit.)

mp

−6 °C (lit.)

density

1.03 g/mL at 25 °C (lit.)
1.034-1.036 g/mL

SMILES string

Cn1ccnc1

InChI

1S/C4H6N2/c1-6-3-2-5-4-6/h2-4H,1H3

InChI key

MCTWTZJPVLRJOU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Methylimidazole is an N-donor ligand generally used in the preparation of coordinate complexes.

Application

1-Methylimidazole may be used as a starting material to synthesize:
  • 1-(2-hydroxypropyl)-3-methylimidazolium salts used as ionic liquids
  • 1,3-dimethylimidazoyl-2-ylidene borane, a stable and easy to handle N-heterocyclic carbene borane
  • It can act both as a catalyst and a solvent in the synthesis of aldononitrile acetates from aldoses.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nickel (II)-, cobalt (II)-, copper (II)-, and zinc (II)-phthalate and 1-methylimidazole coordination compounds: synthesis, crystal structures and magnetic properties.
Baca SG, et al.
Inorgorganica Chimica Acta, 357(12), 3419-3429 (2004)
The use of 1-methylimidazole as a solvent and catalyst for the preparation of aldononitrile acetates of aldoses.
Chen CC & McGinnis GD
Carbohydrate Research, 90(1), 127-130 (1981)
New ionic liquids containing an appended hydroxyl functionality from the atom-efficient, one-pot reaction of 1-methylimidazole and acid with propylene oxide.
Holbrey JD, et al.
Green Chemistry, 5(6), 731-736 (2003)
1,3-Dimethylimidazoyl-2-ylidene borane
Gardner S, et al.
Organic Syntheses, 92, 342-342 (2015)
Fumitoshi Shibahara et al.
The Journal of organic chemistry, 77(19), 8815-8820 (2012-09-15)
Synthetic methods for triarylated azoles containing three different aryl groups via one-pot sequential multiple C-H bond arylations are described. The one-pot sequential diarylation of C5-monoarylated azoles was achieved by the simple sequential addition of two different aryl iodides with a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service