Skip to Content
Merck
All Photos(1)

Key Documents

33497

Supelco

Sarafloxacin hydrochloride hydrate

VETRANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H17F2N3O3 · HCl · xH2O
CAS Number:
Molecular Weight:
421.83 (anhydrous basis)
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Fc1ccc(cc1)N2c3c(cc(c(c3)N4CCNCC4)F)C(=O)C(=C2)C(=O)O.[Cl-].[H+]

InChI

1S/C20H17F2N3O3.ClH/c21-12-1-3-13(4-2-12)25-11-15(20(27)28)19(26)14-9-16(22)18(10-17(14)25)24-7-5-23-6-8-24;/h1-4,9-11,23H,5-8H2,(H,27,28);1H

InChI key

KNWODGJQLCISLC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeong-Heui Choi et al.
Biomedical chromatography : BMC, 25(3), 405-411 (2010-07-06)
In this study, a method for the detection of sarafloxacin in pig and chicken muscles was developed using HPLC-FLD as a regulatory residue technique. Good extraction efficiency was achieved using a mixture of 1% orthophosphoric acid-0.2 m MgCl(2) in water
Patrik Rossi et al.
Intensive care medicine, 34(5), 873-880 (2008-01-25)
To analyze the effect of endothelin-1 on pulmonary arterial and venous contractile force in vitro and on up- and downstream pulmonary vascular resistance in vivo under sham and endotoxemic conditions in pigs. In vitro: paired preparations of pulmonary arteries and
Eric Verdon et al.
Journal of AOAC International, 88(4), 1179-1192 (2005-09-15)
Quinolone antibacterials are veterinary drugs authorized for use in food animal production. The analysis of residual amounts of drugs in food from animal origin is important for quality control of products for consumers. For this purpose, Maximum Residue Limits (MRLs)
Priscilla Henno et al.
American journal of physiology. Lung cellular and molecular physiology, 300(6), L831-L839 (2011-03-08)
Pulmonary vascular remodeling and dysfunction associated to tobacco smoking might pave the way for the subsequent development of pulmonary hypertension. Its prognosis is dreadful and its underlying mechanisms are so far largely unknown in humans. To assess the potential role
Carmen Navarro-Fontestad et al.
Pharmaceutical statistics, 11(1), 14-23 (2011-01-27)
The aim of the present work was to develop a new mathematical method for estimating the area under the curve (AUC) and its variability that could be applied in different preclinical experimental designs and amenable to be implemented in standard

Articles

LC-MS/MS-based determination of 19 sulfonamides, 13 quinolones, and 3 tetracycline drug residues in pork meat samples using Supel™ Swift HLB for extraction.

LC-MS/MS-based determination of 19 sulfonamides, 13 quinolones, and 3 tetracycline drug residues in pork meat samples using Supel™ Swift HLB for extraction.

LC-MS/MS-based determination of 19 sulfonamides, 13 quinolones, and 3 tetracycline drug residues in pork meat samples using Supel™ Swift HLB for extraction.

LC-MS/MS-based determination of 19 sulfonamides, 13 quinolones, and 3 tetracycline drug residues in pork meat samples using Supel™ Swift HLB for extraction.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service