Skip to Content
Merck
All Photos(2)

Key Documents

33493

Supelco

2,6-TDI

analytical standard

Synonym(s):

Tolylene-2,6-diisocyanate, 2-Methyl-m-phenylene diisocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NCO)2
CAS Number:
Molecular Weight:
174.16
Beilstein:
2211546
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

autoignition temp.

>1148 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.571 (lit.)

bp

129-133 °C/18 mmHg (lit.)

density

1.225 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

Cc1c(cccc1N=C=O)N=C=O

InChI

1S/C9H6N2O2/c1-7-8(10-5-12)3-2-4-9(7)11-6-13/h2-4H,1H3

InChI key

RUELTTOHQODFPA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Find all available reference materials for compounds listed in 10/2011 here

Application

2,6-TDI may be used as a reference standard for the quantification of residual 2,6-TDI in foam samples using high performance liquid chromatography coupled to coordination-ionspray tandem mass spectrometry (HPLC-CIS-MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sebastien Gagne et al.
Journal of environmental monitoring : JEM, 7(2), 145-150 (2005-02-04)
Occupational exposures to isocyanates can lead to occupational asthma. Once sensitized, some workers could react to isocyanate monomers at concentrations below 1% of the Permissible Exposure Limit of 5 ppb in air. Currently available methods are not sufficiently sensitive to
L I Maklakov et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 71(1), 238-244 (2008-02-19)
Restricted Hartree-Fock and density function calculations (B3LYP), using 6-311++G(d,p), have been used to investigate the far infrared spectra of aromatic urethanes, synthesized on the basis of 2,4-and 2,6-toluene diisocyanate (2,4-TDI, 2,6-TDI), and the spectrum of ethylphenylurethane. It is shown, that
G Sesana et al.
American Industrial Hygiene Association journal, 52(5), 183-186 (1991-05-01)
A new sampling system is presented that uses solid sorbent media contained in a tube for the determination of airborne isocyanates (2.4-2.6 toluene diisocyanate, hexamethylene diisocyanate, and 4.4' diaminodiphenylmethane diisocyanate). The method is compared with the National Institute for Occupational
Justin M Hettick et al.
Analytical biochemistry, 414(2), 232-238 (2011-04-05)
Diisocyanates are highly reactive chemical compounds widely used in the manufacture of polyurethanes. Although diisocyanates have been identified as causative agents of allergic respiratory diseases, the specific mechanism by which these diseases occur is largely unknown. To better understand the
Hui-Jung Yeh et al.
Journal of applied toxicology : JAT, 28(2), 189-195 (2007-06-22)
Toluene diisocyanates (TDI) are commonly used in polyurethane (PU)-related products. TDIs have been documented as the leading cause of occupational asthma. Skin exposure to TDI in the workplace is common. However, no studies in the literature have investigated the exact

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service