Skip to Content
Merck
All Photos(2)

Key Documents

32492

Supelco

Florfenicol amine

VETRANAL®, analytical standard

Synonym(s):

R)-α-[(1S)-1-Amino-2-fluoroethyl]-4-(methylsulfonyl)benzenemethanol, D-(−)-threo-2-Amino-3-fluoro-1-[4-(methylsulfonyl)phenyl]-1-propanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14FNO3S
CAS Number:
Molecular Weight:
247.29
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CS(=O)(=O)c1ccc(cc1)[C@@H](O)[C@H](N)CF

InChI

1S/C10H14FNO3S/c1-16(14,15)8-4-2-7(3-5-8)10(13)9(12)6-11/h2-5,9-10,13H,6,12H2,1H3/t9-,10-/m1/s1

InChI key

XLSYLQDVLAXIKK-NXEZZACHSA-N

General description

Florfenicol amine is a polar metabolite of florfenicol.

Application

Florfenicol amine may be used as an analytical reference standard for the determination of the analyte in:
  • Poultry and porcine muscle and liver by gas chromatography-negative chemical ionization mass spectrometry (GC-NCI/MS) with selected ion monitoring (SIM) detection.
  • Food samples by ultra-high performance liquid chromatography-electrospray ionization-tandem mass spectrometry (UHPLC-ESI-MS/MS) operating in selected reaction monitoring (SRM) detection mode.
  • Chicken muscle by LC-ESI-MS/MS with multiple reaction monitoring (MRM) detection.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianzhong Shen et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 877(14-15), 1523-1529 (2009-04-28)
A sensitive and reliable method using gas chromatography-negative chemical ionization mass spectrometry (GC-NCI/MS) was developed for the simultaneous determination of chloramphenicol (CAP), thiamphenicol (TAP), florfenicol (FF), and florfenicol amine (FFA) at trace levels in muscle and liver. Before extraction with
Jiancheng Li et al.
Journal of agricultural and food chemistry, 54(25), 9614-9619 (2006-12-07)
A study of the tissue depletion of florfenicol (FF) administered intramuscularly twice to swine at a dose rate of 20 mg per kg of body weight at 24 h intervals was carried out. Forty healthy cross swine were treated with
Christopher L Wrzesinski et al.
Journal of AOAC International, 86(3), 515-520 (2003-07-11)
A method for quantifying florfenicol amine (FFA) in channel catfish muscle was validated according to U.S. Food and Drug Administration guidelines. FFA is the proposed marker residue for the veterinary antibiotic florfenicol in catfish muscle for regulatory surveillance purposes. The
Byung-Kwon Park et al.
Research in veterinary science, 84(1), 85-89 (2007-06-16)
A study on the bioavailability and pharmacokinetics of florfenicol was conducted in six healthy dogs following a single intravenous (i.v.) or oral (p.o.) dose of 20 mg kg(-1) body weight (b.w.). Florfenicol concentrations in serum were determined by a high-performance
B-K Park et al.
Journal of veterinary pharmacology and therapeutics, 29(1), 37-40 (2006-01-20)
The pharmacokinetics of florfenicol and its metabolite, florfenicol amine, was investigated after its intravenous (i.v.) and oral (p.o.) administration of 20 mg/kg of body weight in Korean catfish (Silurus asotus). After i.v. florfenicol injection (as a bolus), the terminal half-life

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service