Skip to Content
Merck
All Photos(3)

Documents

317802

Sigma-Aldrich

4-(4-Isothiocyanatophenylazo)-N,N-dimethylaniline

97%

Synonym(s):

4-(Dimethylamino)azobenzene-4′-isothiocyanate, DABITC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SCNC6H4N=NC6H4N(CH3)2
CAS Number:
Molecular Weight:
282.36
Beilstein:
752568
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Assay

97%

form

crystals

mp

167-171 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CN(C)c1ccc(cc1)\N=N\c2ccc(cc2)N=C=S

InChI

1S/C15H14N4S/c1-19(2)15-9-7-14(8-10-15)18-17-13-5-3-12(4-6-13)16-11-20/h3-10H,1-2H3/b18-17+

InChI key

OSWZKAVBSQAVFI-ISLYRVAYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Kinoshita et al.
Bioscience, biotechnology, and biochemistry, 62(8), 1488-1491 (1998-10-03)
An easy and highly sensitive method for measuring histamine by HPLC analysis coupled with precolumn derivatization was established. The amino group of histamine was completely colorimetrically labelled with 4-N,N-dimethylamino-azobenzene-4'-isothiocyanate (DABITC) in the presence of sodium bicarbonate at 90 degrees C
Evidence for the formation of a novel glutathione conjugate in the metabolism of an aromatic amine derivative.
D H Hutson et al.
Drug metabolism and disposition: the biological fate of chemicals, 12(4), 523-524 (1984-07-01)
J Y Chang
The Journal of biological chemistry, 264(6), 3111-3115 (1989-02-25)
A new water-soluble color reagent, 4-N,N-dimethylaminoazobenzene-4'-isothiocyano-2'-sulfonic acid (S-DABITC), was used to identify lysine residues of antithrombin III which participate in the binding of heparin. Antithrombin, modified with S-DABITC in the presence and absence of low molecular weight heparin (Mr 5000)
M Inoue et al.
Cancer research, 61(3), 950-956 (2001-02-28)
A synthetic peptide corresponding to the human MUC2 tandem repeat unit was glycosylated in vitro using UDP-GalNAc and extracts of colonic adenocarcinoma and paired normal mucosa, followed by fractionation of the products by reverse phase high-performance liquid chromatography. Several peaks
I Jelesarov et al.
European journal of biochemistry, 216(1), 57-66 (1993-08-15)
The chloroplast enzyme ferredoxin: NADP+ reductase (FNR) catalyzes the reduction of NADP+ by ferredoxin (Fd). FNR and Fd form a 1:1 complex that is stabilized by electrostatic interactions between acidic residues of Fd and basic residues of FNR. To localize

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service