Skip to Content
Merck
All Photos(1)

Key Documents

31209

Supelco

Tolclofos-methyl

PESTANAL®, analytical standard

Synonym(s):

O-(2,6-Dichloro-4-methylphenyl) O,O-dimethyl phosphorothioate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11Cl2O3PS
CAS Number:
Molecular Weight:
301.13
Beilstein:
2136521
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COP(=S)(OC)Oc1c(Cl)cc(C)cc1Cl

InChI

1S/C9H11Cl2O3PS/c1-6-4-7(10)9(8(11)5-6)14-15(16,12-2)13-3/h4-5H,1-3H3

InChI key

OBZIQQJJIKNWNO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tolclofos-methyl is a fungicide, effective in controlling Rhizoctonia solani andCorticium rolfsii.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Tolclofos-methyl may be used as a reference standard for the determination of the analyte in fruits and vegetables using atmospheric pressure gas chromatography quadrupole-time-of-flight mass spectrometry.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of organophosphorus pesticides in fruits and vegetables using atmospheric pressure gas chromatography quadrupole-time-of-flight mass spectrometry.
Cheng Z, et al.
Food Chemistry, 231, 365-373 (2017)
Mode of antifungal action of a new fungicide, tolclofos-methyl.
Mode of Action, Metabolism and Toxicology, 153-157 (1983)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service