Skip to Content
Merck
All Photos(2)

Documents

29173

Supelco

Disperse Orange 1

analytical standard

Synonym(s):

4-(4-Nitrophenylazo)diphenylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4N=NC6H4NHC6H5
CAS Number:
Molecular Weight:
318.33
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥96.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

[O-][N+](=O)c1ccc(cc1)\N=N\c2ccc(Nc3ccccc3)cc2

InChI

1S/C18H14N4O2/c23-22(24)18-12-10-17(11-13-18)21-20-16-8-6-15(7-9-16)19-14-4-2-1-3-5-14/h1-13,19H/b21-20+

InChI key

YFVXLROHJBSEDW-QZQOTICOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Disperse Orange 1 is an azo dye characterized by one or more -N≡N- (azo) groups attached to aromatic systems and is primarily used for coloring purposes. Azo dyes are known to have an adverse impact on human and environmental health.

Application

Disperse Orange 1 may be used as a reference standard for the determination of the disperse dye:
  • In textile products by the ultra-high performance supercritical fluid chromatography (UHPSFC) system coupled with triple quadrupole tandem mass spectrometry (MS/MS).
  • In environmental aquatic samples using solid-phase extraction and liquid chromatography coupled to electrospray ionization tandem mass spectrometry (SPE-LC-ESI-MS/MS).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of 17 disperse dyes in textile by ultra-high performance supercritical fluid chromatography combined with tandem mass spectrometry.
Zhou Y, et al.
Talanta, 127(1-2) (2014)
Evaluation of the interactions of DNA with the textile dyes Disperse Orange 1 and Disperse Red 1 and their electrolysis products using an electrochemical biosensor.
Uliana CV, et al.
Sensors and Actuators B, Chemical, 178, 627-635 (2013)
Gareth O S Williams et al.
Lab on a chip, 12(18), 3356-3361 (2012-07-07)
Photonic crystal fibre constitutes an optofluidic system in which light can be efficiently coupled into a solution-phase sample, contained within the hollow core of the fibre, over long path-lengths. This provides an ideal arrangement for the highly sensitive monitoring of
Using SPE-LC-ESI-MS/MS analysis to assess disperse dyes in environmental water samples.
Zocolo GJ, et al.
Journal of Chromatographic Science, 53(8) (2015)
The azo dyes Disperse Red 1 and Disperse Orange 1 increase the micronuclei frequencies in human lymphocytes and in HepG2 cells.
Chequer FMD, et al.
Mutation Research. Genetic Toxicology and Environmental Mutagenesis, 676(1-2), 83-86 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service