Skip to Content
Merck
All Photos(1)

Documents

27089

Supelco

Chromoionophore IV

Selectophore

Synonym(s):

5-Octadecanoyloxy-2-(4-nitrophenylazo)phenol, ETH 2412

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H43N3O5
CAS Number:
Molecular Weight:
525.68
Beilstein:
8244430
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

product line

Selectophore

Quality Level

mp

121-125 °C

SMILES string

OC1=CC(OC(CCCCCCCCCCCCCCCCC)=O)=CC=C1/N=N/C2=CC=C(C=C2)[N+]([O-])=O

InChI

1S/C30H43N3O5/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-30(35)38-27-22-23-28(29(34)24-27)32-31-25-18-20-26(21-19-25)33(36)37/h18-24,34H,2-17H2,1H3/b32-31+

InChI key

OHHICQPUQDFJLP-QNEJGDQOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chromoionophore IV (5-Octadecanoyloxy-2-(4-nitrophenylazo)phenol, ETH 2412) is a lipophilic pH indicator and is generally used during the fabrication of optical sensors.
Visit our Sensor Applications portal to learn more.

Application

ETH 2412 may be used for the optode membrane preparation, for determination of perchlorate spectrophotometrically.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Assay of lipophilic anions, e.g. nitrate, with solvent polymeric optode membrane based on ETH 2412

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polymeric membrane pH electrodes based on electrically charged ionophores.
Yanming M
Analytical Chemistry, 70 (24), 5252-5258 (1998)
Optical ion sensing with immobilized thin films of photocrosslinked decyl methacrylate.
Ambrose TM and Meyerhoff ME.
Analytica Chimica Acta, 378 (1), 119-126 (1999)
Flow-through bulk optode for the spectrophotometric determination of perchlorate.
Soledad G
Microchimica Acta, 143 (1), 59-63 (2003)
P.C. Hauser et al.
Analytical Chemistry, 62, 1919-1919 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service