Skip to Content
Merck
All Photos(1)

Documents

21365

Sigma-Aldrich

(−)-Camphor-10-sulfonic acid

purum, ≥98.0% (T)

Synonym(s):

(1R)-(−)-Camphor-10-sulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O4S
CAS Number:
Molecular Weight:
232.30
Beilstein:
2809676
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥98.0% (T)

optical activity

[α]20/D −21.5±1°, c = 10% in H2O

SMILES string

CC1(C)C2CC[C@]1(CS(O)(=O)=O)C(=O)C2

InChI

1S/C10H16O4S/c1-9(2)7-3-4-10(9,8(11)5-7)6-15(12,13)14/h7H,3-6H2,1-2H3,(H,12,13,14)/t7-,10-/m0/s1

InChI key

MIOPJNTWMNEORI-XVKPBYJWSA-N

Looking for similar products? Visit Product Comparison Guide

Caution

may contain some dark particles

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruizhi Pang et al.
Journal of colloid and interface science, 510, 127-132 (2017-09-25)
Thin film nanocomposite reverse osmosis (TFN RO) membranes incorporated with hydrophilic nanoparticles show a potential problem that the salt rejection can not be improved significantly. In this study, novel TFN RO membranes incorporated with hydrophobic fluorinated silica nanoparticles were fabricated
Kaoru Nobuoka et al.
Physical chemistry chemical physics : PCCP, 9(44), 5891-5896 (2007-11-09)
We directly observe the interaction between 1-butyl-3-methylimidazolium (bmim) or 1-butyl-2,3-dimethylimidazolium (bm(2)im) and the solute, ethyl acrylate (EA), which is the popular dienophile in the Diels-Alder reaction and an H-bonding acceptor, by using specially designed electrospray mass spectrometry. In imidazolium ionic
Chen Liu et al.
Organic letters, 12(10), 2278-2281 (2010-04-24)
The combination of 9-amino-9-deoxy-epi-cinchonine and (+)-CSA resulted in a novel primary amine-based organocatalyst for effective iminium activation of alpha,beta-unsaturated ketones. Such a catalytic system could catalyze the conjugate addition of nitroacetate to enones in a highly enantioselective manner, affording the
A Kumar et al.
Nanotechnology, 21(17), 175102-175102 (2010-04-02)
Polyaniline (PAni) nanofibers doped with HCl and CSA have been irradiated with 90 MeV O(7+) ions with fluence of 3 x 10(10), 3 x 10(11) and 1 x 10(12) ions cm(-2). TEM micrographs show a decrease in the fiber diameter
Anne-Catherine Servais et al.
Electrophoresis, 25(16), 2701-2710 (2004-09-08)
The enantiomeric separation of a series of basic pharmaceuticals (beta-blockers, local anesthetics, sympathomimetics) has been investigated in nonaqueous capillary electrophoresis (NACE) systems using heptakis(2,3-di-O-methyl-6-O-sulfo)-beta-cyclodextrin (HDMS-beta-CD) in combination with potassium camphorsulfonate (camphorSO3-). For this purpose, a face-centered central composite design with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service