Skip to Content
Merck
All Photos(1)

Documents

17118

Millipore

Aeromonas Isolation Agar (Base)

suitable for microbiology, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171609
NACRES:
NA.85

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 12.5 g/L
L-arginine hydrochloride, 2 g/L
bile salts, 3 g/L
bromothymol blue, 0.04 g/L
ferric ammonium citrate, 0.8 g/L
inositol, 2.5 g/L
lactose, 1.5 g/L
L-lysine hydrochloride, 3.5 g/L
peptone, special, 5 g/L
sodium chloride, 5 g/L
sodium thiosulfate, 10.67 g/L
sorbose, 3 g/L
thymol blue, 0.04 g/L
xylose, 3.75 g/L
yeast extract, 3 g/L

manufacturer/tradename

NutriSelect® Plus

final pH

8.0±0.2 (25 °C)

application(s)

agriculture
clinical testing
environmental
food and beverages

microbiology

suitability

Aeromonas spp.

Application

Used for the selective, differential isolation of Aeromonas hydrophila from clinical and environmental specimens.

Preparation Note

Suspend 28.15 g in 500 ml distilled water. Heat to boiling to dissolve the medium completely. Do not autoclave. Cool to 50°C and aseptically add rehydrated contents of 1 vial of Aeromonas Selective Supplement (Cat. No. No. 17119). Mix well and dispense for use as desired.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service