Skip to Content
Merck
All Photos(2)

Key Documents

06957

Supelco

(−)-Chicoric acid

analytical standard

Synonym(s):

(2R,3R)-2,3-Bis{[(2E)-3-(3,4-dihydroxyphenyl)-1-oxo-2-propen-1-yl]oxy}butanedioic acid, 2,3-Di-trans-caffeoyltartaric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H18O12
CAS Number:
Molecular Weight:
474.37
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)[C@H](OC(=O)\C=C\c1ccc(O)c(O)c1)[C@@H](OC(=O)\C=C\c2ccc(O)c(O)c2)C(O)=O

InChI

1S/C22H18O12/c23-13-5-1-11(9-15(13)25)3-7-17(27)33-19(21(29)30)20(22(31)32)34-18(28)8-4-12-2-6-14(24)16(26)10-12/h1-10,19-20,23-26H,(H,29,30)(H,31,32)/b7-3+,8-4+/t19-,20-/m1/s1

InChI key

YDDGKXBLOXEEMN-IABMMNSOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chicoric acid, a caffeic acid derivative, is found in a variety of plant species, including basil, chicory, dandelion, etc. It is a polyphenol organic compound that is reported to have antioxidant, anti-inflammatory, antiviral and immunostimulating properties. Various studies also suggest the antidiabetic potential of chicoric acid.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: echinacea

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stacey N Barnaby et al.
Nanotechnology, 23(29), 294011-294011 (2012-06-30)
In this work, we report the formation of well-defined silver nanodendrites via biomineralization under mild conditions in a single step, in the presence of the plant phytohormone chicoric acid (CA), a well-known HIV-I integrase inhibitor. CA played a dual role
Narimane Segueni et al.
Planta medica, 77(10), 999-1004 (2011-02-10)
Stromelysin-1 (matrix metalloproteinase-3: MMP-3) occupies a central position in collagenolytic and elastolytic cascades, leading to cutaneous intrinsic and extrinsic aging. We screened extracts of a propolis sample from Algeria with the aim to isolate compounds able to selectively inhibit this
Didier Tousch et al.
Biochemical and biophysical research communications, 377(1), 131-135 (2008-10-07)
Caffeic acid and chlorogenic acid (CGA), a mono-caffeoyl ester, have been described as potential antidiabetic agents. Using in vitro studies, we report the effects of a dicaffeoyl ester, chicoric acid (CRA) purified from Cichorium intybus, on glucose uptake and insulin
Rachid Bel-Rhlid et al.
Journal of agricultural and food chemistry, 60(36), 9236-9241 (2012-08-28)
Chicoric acid (ChA) and caftaric acid (CafA) were identified as bioactive components of chicory and have been ascribed a number of health benefits. This study investigated the hydrolysis of ChA and CafA with enzymes and a probiotic bacterium Lactobacillus johnsonii
Chung Mu Park et al.
Journal of ethnopharmacology, 133(2), 834-842 (2010-11-16)
The common dandelion (Taraxacum officinale G.H. Weber ex Wiggers, Asteraceae) has been widely used in folklore medicine to treat dyspepsia, heartburn, and spleen and liver disorders. To compare the antioxidative and anti-inflammatory activities of Taraxacum officinale methanol extract (TOME) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service