Skip to Content
Merck
All Photos(1)

Key Documents

04238

Sigma-Aldrich

Hydroxyapatite

puriss., meets analytical specification of Ph. Eur., BP, FCC, E341, ≥90% (calculated on glowed substance)

Synonym(s):

Calcium phosphate hydroxide, Durapatite, Hydroxylapatite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
3Ca3(PO4)2 · Ca(OH)2
CAS Number:
Molecular Weight:
502.31
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.21

grade

puriss.

Quality Level

Assay

≥90% (calculated on glowed substance)

form

powder

quality

meets analytical specification of Ph. Eur., BP, FCC, E341

composition

Ca, 35-40%

impurities

residual solvents, complies
≤0.001% heavy metals (as Pb)
≤0.2% insoluble in HCl

loss

≤8% loss on ignition, 800 °C

anion traces

chloride (Cl-): ≤1500 mg/kg
fluoride (F-): ≤50 mg/kg
sulfate (SO42-): ≤5000 mg/kg

cation traces

As: ≤2 mg/kg
Cd: ≤1 mg/kg
Cu: ≤20 mg/kg
Fe: ≤400 mg/kg
Hg: ≤1 mg/kg
Pb: ≤2 mg/kg
Zn: ≤20 mg/kg

SMILES string

[Ca++].[Ca++].[Ca++].[Ca++].O[Ca+].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O

InChI

1S/5Ca.3H3O4P.H2O/c;;;;;3*1-5(2,3)4;/h;;;;;3*(H3,1,2,3,4);1H2/q5*+2;;;;/p-10

InChI key

XYJRXVWERLGGKC-UHFFFAOYSA-D

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • New Physico-Chemical Analysis of Magnesium-Doped Hydroxyapatite in Dextran Matrix Nanocomposites.: This research article focuses on the development and characterization of magnesium-doped hydroxyapatite within a dextran matrix, emphasizing its potential applications in biomedical and tissue engineering fields (Predoi et al., 2023).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mechanical properties of sintered hydroxyapatite for prosthetic applications.
Akao M ,et al.
J. Mater. Sci., 16(3), 809-812 (1981)
P Ducheyne et al.
Biomaterials, 7(2), 97-103 (1986-03-01)
Hydroxyapatite from two sources was electrophoretically deposited onto flat titanium plate material. Depending upon the deposition conditions various changes in the structure of the ceramic were identified. A well-adhering Ti-P compound was present at the interface. Hydroxyapatite oxygenated to various
Chemical characterization of silicon-substituted hydroxyapatite
Gibson I, et al.
Journal of Biomedical Materials Research, 44(4), 422-428 (1999)
Sophie E P New et al.
Circulation research, 113(1), 72-77 (2013-04-26)
We previously showed that early calcification of atherosclerotic plaques associates with macrophage accumulation. Chronic renal disease and mineral imbalance accelerate calcification and the subsequent release of matrix vesicles (MVs), precursors of microcalcification. We tested the hypothesis that macrophage-derived MVs contribute
Bjarke Viberg et al.
Acta orthopaedica, 84(3), 254-259 (2013-04-19)
Elderly patients with displaced femoral neck fractures are commonly treated with a hemiarthroplasty (HA), but little is known about the long-term failure of this treatment. We compared reoperation rates for patients aged at least 75 years with displaced femoral neck

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service