Skip to Content
Merck
All Photos(1)

Key Documents

03918

Supelco

DL-Tartaric acid

anhydrous, suitable for ion chromatography, ≥99.5% (T)

Synonym(s):

DL-2,3-Dihydroxybutanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOC(CHOH)2COOH
CAS Number:
Molecular Weight:
150.09
Beilstein:
1725148
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NB.21

Quality Level

description

anionic

Assay

≥99.5% (T)

shelf life

limited shelf life, expiry date on the label

technique(s)

ion chromatography: suitable

mp

210-212 °C (lit.)

solubility

H2O: 0.1 g/mL, clear

cation traces

Al: ≤5 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤10 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
NH4+: ≤10 mg/kg
Na: ≤10 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

O[C@@H]([C@H](O)C(O)=O)C(O)=O

InChI

1S/C4H6O6/c5-1(3(7)8)2(6)4(9)10/h1-2,5-6H,(H,7,8)(H,9,10)/t1-,2-/m0/s1

InChI key

FEWJPZIEWOKRBE-LWMBPPNESA-N

Looking for similar products? Visit Product Comparison Guide

Application

Metrohm IC application note AN-C-131: Magnesium,cadmium, and iron in phosphoric acid.

Metrohm IC application note AN-C-111:N-Methylpyrrolidone impurity in Cefepime

Metrohm IC application note AN-C-100: Choline ininfant milk powder using online dialysis.

Linkage

Visit the IC Portal to learn more

related product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

410.0 °F - closed cup

Flash Point(C)

210 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mrinal Kanti Bain et al.
Carbohydrate polymers, 91(2), 529-536 (2012-11-06)
Gelation temperature of MC was reduced from 59°C to 54°C with the addition of 10% PEG. Sodium tartrate (NaT) and sodium citrate (NaC) were added to the MC-PEG solution to further reduce the gelation temperature close to physiological temperature. Different
J B Olivato et al.
Carbohydrate polymers, 92(2), 1705-1710 (2013-02-13)
Tartaric acid (TA), a dicarboxylic acid, can act as a compatibiliser in starch/polyester blends. A mixture design was proposed to evaluate the effect of TA on the properties of starch/poly (butylene adipate co-terephthalate) (PBAT) blown films plasticised with glycerol. The
Mark D Eddleston et al.
Chemical communications (Cambridge, England), 48(92), 11340-11342 (2012-10-18)
The formation of diastereomeric cocrystals of malic acid and tartaric acid was investigated by liquid-assisted grinding in the solid state. We demonstrate that racemic malic acid can be converted into two distinct diastereomeric cocrystal phases by grinding with a single
Jianping Yang et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(43), 13642-13650 (2012-09-22)
In this paper, we report a facile one-step hydrothermal method to synthesize phase-, size-, and shape-controlled carboxyl-functionalized rare-earth fluorescence upconversion phosphors by using a small-molecule binary acid, such as malonic acid, oxalic acid, succinic acid, or tartaric acid as capping
Carmen-Alice Teacă et al.
Carbohydrate polymers, 93(1), 307-315 (2013-03-08)
The present paper describes the preparation and characterization of polysaccharides-based bio-composite films obtained by the incorporation of 10, 20 and 30 wt% birch cellulose (BC) within a glycerol plasticized matrix constituted by the corn starch (S) and chemical modified starch

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service