Skip to Content
Merck
All Photos(1)

Key Documents

03820585

Isoorientin

primary reference standard

Synonym(s):

Homoorientin, Luteolin 6-C-β-D-glucoside, Luteolin 6-C-glucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20O11
CAS Number:
Molecular Weight:
448.38
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

application(s)

food and beverages

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)c2c(O)cc3OC(=CC(=O)c3c2O)c4ccc(O)c(O)c4

InChI

1S/C21H20O11/c22-6-14-17(27)19(29)20(30)21(32-14)16-11(26)5-13-15(18(16)28)10(25)4-12(31-13)7-1-2-8(23)9(24)3-7/h1-5,14,17,19-24,26-30H,6H2/t14-,17-,19+,20-,21+/m1/s1

InChI key

ODBRNZZJSYPIDI-VJXVFPJBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference Standard in the analysis of herbal medicinal products

Biochem/physiol Actions

C-glycosyl flavone with anti-inflammatory, antimicrobial and antioxidant properties. Induces antioxidant response through PI3K singaling.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li Yuan et al.
Toxicology and applied pharmacology, 265(1), 83-92 (2012-10-03)
Isoorientin (ISO) is a flavonoid compound that can be extracted from several plant species, such as Phyllostachys pubescens, Patrinia, and Drosophyllum lusitanicum; however, its biological activity remains poorly understood. The present study investigated the effects and putative mechanism of apoptosis
Roxana Orrego et al.
Molecules (Basel, Switzerland), 14(10), 3906-3913 (2009-11-20)
This study assessed the inhibitory effect of three C-glycosylflavonoids from Cymbopogon citratus leaves--isoorientin (1), swertiajaponin (2) and isoorientin 2"-Orhamnoside (3)--on human LDL oxidation. Isolated LDL was incubated with compounds 1-3 and the kinetics of lipid peroxidation were assessed by conjugated
Severina Pacifico et al.
Journal of natural products, 73(12), 1973-1978 (2010-11-18)
Eight flavonoid C-glycosides, including three new analogues, have been isolated from leaf and root methanolic extracts of Petrorhagia velutina, a Mediterranean herbaceous plant. The antiproliferative activity against human hepatoblastoma cancer cell line HepG2 has been analyzed by the MTT (3-(4,5-dimethyl-2-thiazolyl)-2,5-diphenyl-2H-tetrazolium
Dong-Seon Kim et al.
Planta medica, 76(7), 678-682 (2009-12-05)
We aimed to find antiallergic agents from natural sources using mast cells activated during allergic reaction. We screened over 2000 plants for blockade of histamine release and identified two of them, S. baicalenesis and P. edulis. Bioassay-guided fractionation led to
Jinyong Peng et al.
Journal of chromatography. A, 1074(1-2), 111-115 (2005-06-09)
High-speed counter-current chromatography (HSCCC) with a solvent system composed of ethyl acetate-n-butanol-water (2:1:3, v/v/v) was used to isolate and separate two C-glycosylflavones from Patrinia villosa Juss, a traditional Chinese medicine. The separation produced 42.9 mg isovitexin and 20.1 mg isoorientin

Protocols

HPTLC fingerprinting enables rapid identification of passion flower with related reference materials.

HPTLC fingerprinting enables rapid identification of passion flower with related reference materials.

HPTLC fingerprinting enables rapid identification of passion flower with related reference materials.

HPTLC fingerprinting enables rapid identification of passion flower with related reference materials.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service