Skip to Content
Merck
All Photos(2)

Key Documents

03813

Supelco

Dihydrocapsaicin

analytical standard

Synonym(s):

6,7-Dihydrocapsaicin, 8-Methyl-N-vanillylnonanamide, N-[(4-Hydroxy-3-methoxyphenyl)methyl]-8-methylnonenamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H29NO3
CAS Number:
Molecular Weight:
307.43
Beilstein:
2815150
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

COc1cc(CNC(=O)CCCCCCC(C)C)ccc1O

InChI

1S/C18H29NO3/c1-14(2)8-6-4-5-7-9-18(21)19-13-15-10-11-16(20)17(12-15)22-3/h10-12,14,20H,4-9,13H2,1-3H3,(H,19,21)

InChI key

XJQPQKLURWNAAH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dihydrocapsaicin belongs to the capsaicinoid group of compounds which are responsible for the pungency of capsicum fruits.

Application

Dihydrocapsaicin has been used as a reference standard for the identification of dihydrocapsaicin in blood and tissue samples by high performance liquid chromatography (HPLC) combined with tandem mass spectrometry (MS) and in tomato-based salsas by enzyme immunoassay (EIA) and LC with fluorescent detection.
It may be used as a reference standard for the determination of dihydrocapsaicin in Capsicum fruit samples by HPLC equipped with a Surveyor photodiode array (PDA) detector and in rat plasma by HPLC-triple quadrupole MS with electrospray ionization (ESI) in selected reaction monitoring (SRM) mode.

Biochem/physiol Actions

VR1 vanilloid receptor agonist.

Linkage

Capsaicin analog.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of capsaicinoids in salsa by liquid chromatography and enzyme immunoassay.
Perkins, et al.
Journal of AOAC (Association of Official Analytical Chemists) International, 85(1), 82-85 (2002)
Optimization of high-performance liquid chromatographic parameters for the determination of capsaicinoid compounds using the simplex method.
Karnka, et al.
Analytical Sciences, 18(6), 661-665 (2002)
Determination of capsaicin, nonivamide, and dihydrocapsaicin in blood and tissue by liquid chromatography-tandem mass spectrometry.
Reilly, et al.
Journal of Analytical Toxicology, 26(6), 313-319 (2002)
Boyeon Park et al.
Journal of microbiology and biotechnology, 29(10), 1580-1590 (2019-09-03)
Capsaicinoids in red pepper powder are known to show anti-bacterial effects; however, their effects during kimchi fermentation are not known. This study aimed to investigate the effects of various concentrations of capsaicinoids on kimchi fermentation. Five sets of kimchi samples
Xinli Mao et al.
Molecular medicine reports, 17(4), 6116-6121 (2018-02-14)
Capsaicin is a principal component of hot red peppers and chili peppers. Previous studies have reported that capsaicin exhibits antitumor functions in a variety of tumor models. Although various mechanisms underlying the capsaicin‑mediated inhibition of tumor growth have been demonstrated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service