Skip to Content
Merck
All Photos(2)

Key Documents

03738

Supelco

5-Fluorouracil

analytical standard

Synonym(s):

2,4-Dihydroxy-5-fluoropyrimidine, 5-FU, 5-Fluoro-2,4(1H,3H)-pyrimidinedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3FN2O2
CAS Number:
Molecular Weight:
130.08
Beilstein:
127172
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

mp

282-286 °C (dec.) (lit.)

application(s)

cleaning products
cosmetics
food and beverages
forensics and toxicology
personal care
pharmaceutical (small molecule)

format

neat

SMILES string

FC1=CNC(=O)NC1=O

InChI

1S/C4H3FN2O2/c5-2-1-6-4(9)7-3(2)8/h1H,(H2,6,7,8,9)

InChI key

GHASVSINZRGABV-UHFFFAOYSA-N

Gene Information

human ... TYMS(7298)

Looking for similar products? Visit Product Comparison Guide

General description

5-Fluorouracil is an anti-tumor agent, widely used in the treatment of various malignancies.

Application

5-Fluorouracil may be used as an analytical standard for the determination of the analyte in hospital effluents by capillary electrophoresis, and in biological samples and environmental samples by chromatography based techniques.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A sensitive method for determination of 5-fluorouracil and 5-fluoro-2'-deoxyuridine in human plasma by high-pressure liquid chromatography.
A R Buckpitt et al.
Analytical biochemistry, 106(2), 432-437 (1980-08-01)
5-fluorouracil: mechanisms of action and clinical strategies
Longley DB, et al.
Nature Reviews. Cancer, 3(5), 330-338 (2003)
G Micoli et al.
Journal of chromatography. B, Biomedical sciences and applications, 750(1), 25-32 (2001-02-24)
5-Fluorouracil (5-FU) is one of the most widely used antineoplastic drugs. It can be therefore considered to be a model compound for the identification of exposure routes during preparation and administration of cytostatic agents, especially for nucleoside analogue drugs. In
Use of 5-fluorouracil and survival in patients with microsatellite-unstable colorectal cancer
Carethers JM, et al.
Gastroenterology, 126(2), 394-401 (2004)
Susanne N Mahnik et al.
Analytical and bioanalytical chemistry, 380(1), 31-35 (2004-09-15)
Cytostatic anticancer drugs are an increasingly important issue in the environmental debate, mainly due to the lack of knowledge about the fate of these toxic substances. Over the last decades, 5-fluorouracil (5-FU) has been one of the most frequently used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service