Skip to Content
Merck
All Photos(1)

Key Documents

03600585

Luteolin 7-glucoside

primary reference standard

Synonym(s):

Luteolin 7-O-β-D-glucoside, 3′,4′,5,7-Tetrahydroxyflavone 7-glucoside, Cynaroside, Glucoluteolin, Glucosylluteolin, Luteoloside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20O11
CAS Number:
Molecular Weight:
448.38
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

application(s)

food and beverages

SMILES string

OC[C@H]1O[C@@H](Oc2cc(O)c3C(=O)C=C(Oc3c2)c4ccc(O)c(O)c4)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C21H20O11/c22-7-16-18(27)19(28)20(29)21(32-16)30-9-4-12(25)17-13(26)6-14(31-15(17)5-9)8-1-2-10(23)11(24)3-8/h1-6,16,18-25,27-29H,7H2/t16-,18-,19+,20-,21-/m1/s1

InChI key

PEFNSGRTCBGNAN-QNDFHXLGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference Standard in the analysis of herbal medicinal products

Other Notes

This compound is commonly found in plants of the genus: achillea crataegus humulus hypericum melissa mentha pimpinella salvia thymus

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Donatella Negro et al.
Journal of food science, 77(2), C244-C252 (2012-01-19)
Polyphenol compounds, particularly caffeoylquinic acids and flavonoids, were measured in different tissues and developmental stages of 6 artichoke varietal types diffused in the Mediterranean region. Flower heads were subdivided into external, intermediate, internal bracts, and receptacle, while leaves were collected
Rameshwar U Kadam et al.
Journal of medicinal chemistry, 50(25), 6337-6342 (2007-11-09)
The target for the anti-inflammatory natural products like amentoflavone ( 2), which act by interfering with the proinflammatory cytokine pathway (e.g., TNF-alpha, IL-1beta, and NO synthase), is not yet well-defined. Data obtained from docking, electronic, and surface analyses shed some
Ipek Süntar et al.
Journal of ethnopharmacology, 141(3), 1058-1070 (2012-04-24)
In Turkish traditional medicine, the aerial parts of Daphne oleoides Schreber subsp. kurdica (DOK) have been used to treat malaria, rheumatism and for wound healing. The aim was to evaluate the ethnopharmacological usage of the plant using in vivo and
Liancai Zhu et al.
Chemistry & biodiversity, 6(10), 1716-1726 (2009-10-21)
Agrimonia pilosa Ledeb is used as the tonic for asthenia and fatigue in China. Considering that the energizing effect might be correlated with antioxidant properties, we investigated the antioxidant activities of aqueous extract (AE) from Agrimonia pilosa Ledeb by assessing
Nadine Backhouse et al.
Journal of ethnopharmacology, 119(1), 160-165 (2008-07-22)
Leaf extracts of Buddleja globosa (Buddlejaceae) are used in Chilean folk medicine for wound healing. The anti-inflammatory (topic and per os), analgesic (per os) effects and the antioxidant activity of Buddleja globosa were for the first time reported by us.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service