Skip to Content
Merck
All Photos(3)

Key Documents

02851

Sigma-Aldrich

Ethanol

greener alternative

for residue analysis

Synonym(s):

Ethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07
Beilstein:
1718733
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.05
grade:
for residue analysis
technique(s):
GC/MS: suitable
application(s):
food and beverages
bp:
78 °C (lit.)
78 °C
vapor pressure:
44.6 mmHg ( 20 °C)
44.6 mmHg

grade

for residue analysis

Quality Level

100
500

vapor density

1.59 (vs air)
1.59 (vs air)

vapor pressure

44.6 mmHg ( 20 °C)
44.6 mmHg

product line

PESTANAL®

form

liquid

autoignition temp.

683 °F

expl. lim.

19 %, 60 °F
19 %

greener alternative product characteristics

Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

concentration

≥99.8 % (v/v)

technique(s)

GC/MS: suitable

impurities

≤0.0005% non volatile
≤0.05% water (Karl Fischer)

evapn. residue

≤0.0005%

refractive index

n20/D 1.3600 (lit.)
n20/D 1.361-1.363

bp

78 °C (lit.)
78 °C

mp

-114 °C
−114 °C (lit.)

application(s)

food and beverages

format

neat

greener alternative category

SMILES string

CCO

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the  four categories of Greener Alternatives. This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Other Notes

Sales restrictions may apply

Footnote

1Distribution and use of ethyl alcohol in Switzerland require a license from the Swiss Federal Alcohol Administration

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

also commonly purchased with this product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cem Kuscu et al.
Nature biotechnology, 32(7), 677-683 (2014-05-20)
RNA-guided genome editing with the CRISPR-Cas9 system has great potential for basic and clinical research, but the determinants of targeting specificity and the extent of off-target cleavage remain insufficiently understood. Using chromatin immunoprecipitation and high-throughput sequencing (ChIP-seq), we mapped genome-wide
Stefan Wieland et al.
Hepatology (Baltimore, Md.), 59(6), 2121-2130 (2013-10-15)
Approximately 50% of patients with chronic hepatitis C (CHC) have ongoing expression of interferon stimulated genes (ISGs) in the liver. It is unclear why this endogenous antiviral response is inefficient in eradicating the infection. Several viral escape strategies have been
S Enocksson et al.
The Journal of clinical investigation, 95(5), 2239-2245 (1995-05-01)
Although it is well established in several mammalian species that beta 3-adrenoceptors play a major role in regulating lipolysis and thermogenesis in adipose tissue, the functional existence and role of this receptor subtype in man has been controversial. We investigated
Cecilia E Hanzel et al.
Neurobiology of aging, 35(10), 2249-2262 (2014-05-17)
Chronic brain inflammation is associated with Alzheimer's disease (AD) and is classically attributed to amyloid plaque deposition. However, whether the amyloid pathology can trigger early inflammatory processes before plaque deposition remains a matter of debate. To address the possibility that
Gian Marco Leggio et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 39(8), 2017-2028 (2014-03-04)
Mesolimbic dopamine (DA) controls drug- and alcohol-seeking behavior, but the role of specific DA receptor subtypes is unclear. We tested the hypothesis that D3R gene deletion or the D3R pharmacological blockade inhibits ethanol preference in mice. D3R-deficient mice (D3R(-/-)) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service