Skip to Content
Merck
All Photos(1)

Key Documents

02676

Sigma-Aldrich

N-Methyl-L-alanine

≥98.0% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9NO2
CAS Number:
Molecular Weight:
103.12
Beilstein:
1720922
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (TLC)

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

color

colorless to white

application(s)

peptide synthesis

SMILES string

CN[C@@H](C)C(O)=O

InChI

1S/C4H9NO2/c1-3(5-2)4(6)7/h3,5H,1-2H3,(H,6,7)/t3-/m0/s1

InChI key

GDFAOVXKHJXLEI-VKHMYHEASA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Z Grzonka et al.
International journal of peptide and protein research, 25(4), 375-381 (1985-04-01)
The 600 MHz proton n.m.r. spectra of (sarcosyl7)-oxytocin and (N-methylalanyl7) oxytocin in 2H2O solution have been recorded and completely assigned. In each case the spectrum indicates the presence of two slowly interconverting conformers, which are the cis-trans isomers about the
C H Tan et al.
Biochemical pharmacology, 39(5), 955-958 (1990-03-01)
Synaptosomes isolated from adult rat cerebral cortices were used for studying the uptake of L-leucine by the Na(+)-dependent route. Three non-metabolizable amino acid analogues, which had been used previously to discriminate the Na(+)-dependent A-type uptake system of animal cells, were
D Gazis et al.
International journal of peptide and protein research, 23(1), 78-83 (1984-01-01)
Substituting sarcosine or N-methylalanine for proline in the inhibitory vasopressin analogs dPAVP and d(CH2)5AVP had the following effects: 1) milk ejection and antidiuretic activities were severely depressed, 2) pressor antagonism was maintained but weakened somewhat, and 3) antagonism in the
Zijuan Lai et al.
Nature methods, 15(1), 53-56 (2017-11-28)
Novel metabolites distinct from canonical pathways can be identified through the integration of three cheminformatics tools: BinVestigate, which queries the BinBase gas chromatography-mass spectrometry (GC-MS) metabolome database to match unknowns with biological metadata across over 110,000 samples; MS-DIAL 2.0, a
K Lakshminarayana et al.
Indian journal of experimental biology, 38(4), 373-378 (2001-02-24)
Spontaneous mutants resistant to methionine sulfoximine (Msx), methyl alanine (Mal) and methyl ammonium chloride (Mac) were derived from A. chroococcum strain A103. Msx and Mal-resistant mutants expressed 1.73 to 10.98% of the fully derepressed nitrogenase activity when grown in Burk's

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service