Skip to Content
Merck
All Photos(1)

Key Documents

00990

Sigma-Aldrich

Acetyl chloride

puriss. p.a., ≥99.0% (T)

Synonym(s):

Acetic acid chloride, Acetic chloride, Ethanoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCl
CAS Number:
Molecular Weight:
78.50
Beilstein:
605303
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39050501
PubChem Substance ID:
NACRES:
NA.21

vapor density

2.7 (vs air)

Quality Level

vapor pressure

11.69 psi ( 20 °C)
32.33 psi ( 55 °C)

grade

puriss. p.a.

Assay

≥99.0% (T)

form

liquid

autoignition temp.

1353 °F

expl. lim.

19 %

refractive index

n20/D 1.389 (lit.)
n20/D 1.389

bp

52 °C (lit.)

mp

−112 °C (lit.)

density

1.104 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.1 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.5 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

functional group

acyl chloride

SMILES string

CC(Cl)=O

InChI

1S/C2H3ClO/c1-2(3)4/h1H3

InChI key

WETWJCDKMRHUPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetyl chloride (acyl chloride) is an acetylation reagent used in esterification and Friedel-Crafts acylation reactions.

Application

Acetyl chloride can be used as:
  • A degradation agent in the degradation of polyoxyethylene glycol monoalkyl ethers to determine the degree of ethoxylation using chromatography-mass spectrometry (MS).
  • A reagent with alcohols for the esterification of carboxylic acids, N-boc deprotection, and phosphoramide solvolysis reactions.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

41.0 °F - closed cup

Flash Point(C)

5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparisons of O-acylation and Friedel-Crafts acylation of phenols and acyl chlorides and Fries rearrangement of phenyl esters in trifluoromethanesulfonic acid: Effective synthesis of optically active homotyrosines
Murashige R, et al.
Tetrahedron, 67(3), 641-649 (2011)
A rapid and simple method for the esterification of fatty acids and steroid carboxylic acids prior to gas-liquid chromatography
Lillington JM, et al.
Clinica Chimica Acta; International Journal of Clinical Chemistry, 111(1), 91-98 (1981)
Uta Ceglarek et al.
Metabolomics : Official journal of the Metabolomic Society, 12(12), 182-182 (2016-11-15)
Sensitive and specific assessment of the hepatic graft metabolism after liver transplantation (LTX) is essential for early detection of postoperative dysfunction implying the need for consecutive therapeutic interventions. Here, we assessed circulating liver metabolites of the cholesterol pathway, amino acids
Jacob Bart et al.
Journal of the American Chemical Society, 131(14), 5014-5015 (2009-03-27)
A microfluidic high-resolution NMR flow probe based on a novel stripline detector chip is demonstrated. This tool is invaluable for the in situ monitoring of reactions performed in microreactors. As an example, the acetylation of benzyl alcohol with acetyl chloride
Jennifer L Frahm et al.
Analytical chemistry, 79(11), 3989-3995 (2007-05-05)
The wide range of protein concentrations found in biological matrixes presents a formidable analytical challenge in proteomics experiments. It is predicted that low-abundance proteins are the likely clinically relevant targets in disease-based proteomics analyses. To effectively analyze low-abundance proteins by

Articles

Development and validation of a fatty acid methyl esterification method, using methanolic HCl reagent, for FAME analysis by GC of vegetable oils and fats.

Development and validation of a fatty acid methyl esterification method, using methanolic HCl reagent, for FAME analysis by GC of vegetable oils and fats.

Development and validation of a fatty acid methyl esterification method, using methanolic HCl reagent, for FAME analysis by GC of vegetable oils and fats.

Development and validation of a fatty acid methyl esterification method, using methanolic HCl reagent, for FAME analysis by GC of vegetable oils and fats.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service