Skip to Content
Merck
All Photos(2)

Documents

00881

Supelco

(±)-4-Hydroxypropranolol hydrochloride

analytical standard

Synonym(s):

1-[(4-Hydroxy-1-naphthyl)oxy]-3-(isopropylamino)- 2-propanol hydrochloride, 4′-Hydroxypropranolol hydrochloride, 4-{2-Hydroxy-3-[(1-methylethyl)amino]propoxy}-1-naphthalenol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H21NO3·HCl
CAS Number:
Molecular Weight:
311.80
Beilstein:
4036845
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (AT)
≥98.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(C)NCC(O)COC1=CC=C(O)C2=CC=CC=C21.Cl

InChI

1S/C16H21NO3.ClH/c1-11(2)17-9-12(18)10-20-16-8-7-15(19)13-5-3-4-6-14(13)16;/h3-8,11-12,17-19H,9-10H2,1-2H3;1H

InChI key

ROUJENUXWIFONU-UHFFFAOYSA-N

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T D Eller et al.
Journal of chromatography, 612(2), 320-325 (1993-02-26)
Selective extraction of sulfate conjugates of basic drugs from biological matrices has been difficult because of their highly polar nature. Immunoaffinity isolation may be the best solution to this analytical problem. This was tested for a model compound, the metabolite
Junko Miyano et al.
Biochemical pharmacology, 69(6), 941-950 (2005-03-08)
Pretreatment of Hep G2 cells with beta-naphthoflavone (BNF 1-25microM) significantly increased cytosolic sulfation activities of 4-hydroxypropranolol (4-OH-PL) racemate. The profile was similar to those of sulfations towards dopamine and triiodothyronine in the same cytosolic fractions. Kinetic studies of 4-OH-PL sulfation
Pankaj Partani et al.
Journal of pharmaceutical and biomedical analysis, 50(5), 966-976 (2009-07-31)
A very sensitive, reliable, reproducible and highly selective assay for the simultaneous determination of free and total (conjugated and unconjugated) propranolol and its equipotent hydroxyl metabolite, 4-hydroxy propranolol, in human plasma was developed and validated. The analytes were simultaneously extracted
V G Belolipetskaia et al.
Eksperimental'naia i klinicheskaia farmakologiia, 58(6), 56-59 (1995-11-01)
Pharmacokinetics of propranolol (P), 4-hydroxy-propranolol sulfate (4HOP-Sulf), and glucoronides of pharmacologically active S-enantiomer P (S-PG) and ballast R-enantiomer of P (R-PG) in the blood serum of 21 patients with chronic ischemic heart disease and/or arterial hypertension has been studied at
Takeshi Shimizudani et al.
Chemico-biological interactions, 183(1), 67-78 (2009-10-27)
Oxidative metabolism of propranolol (PL) enantiomers (R-PL and S-PL) to 4-hydroxypropranolol (4-OH-PL), 5-OH-PL and N-deisopropylpropranolol (NDP) was examined in hepatic microsomes from cynomolgus and marmoset monkeys and in small intestinal microsomes from monkeys and humans. In hepatic microsomes, levels of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service