Skip to Content
Merck
All Photos(1)

Key Documents

30-5350

Sigma-Aldrich

Tuerk′s reagent

Synonym(s):

Tuerk solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12000000
form:
liquid

form

liquid

availability

available only in Japan

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

Suitability

leukocytometry

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service