Skip to Content
Merck
All Photos(1)

Key Documents

28-1830

Sigma-Aldrich

Sodium benzoate

SAJ special grade, ≥99.0%

Synonym(s):

Benzoic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COONa
CAS Number:
Molecular Weight:
144.10
Beilstein:
3572467
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
SAJ special grade
form:
solid

grade

SAJ special grade

vapor pressure

<0.01 hPa ( 20 °C)

Assay

≥99.0%

form

solid

availability

available only in Japan

pH

7.0-8.5 (25 °C, 144.1 g/L)

mp

>300 °C (lit.)

storage temp.

15-25°C

SMILES string

[Na+].[O-]C(=O)c1ccccc1

InChI

1S/C7H6O2.Na/c8-7(9)6-4-2-1-3-5-6;/h1-5H,(H,8,9);/q;+1/p-1

InChI key

WXMKPNITSTVMEF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomás Subils et al.
Journal of food protection, 75(5), 959-965 (2012-05-09)
Toxin synthesis by Shiga toxin-producing Escherichia coli (STEC) appears to be coregulated through the induction of the integrated bacteriophages that encode the toxin genes. These phages might be the principal means for the dissemination and release of Shiga toxins. We
Jia Li et al.
Environmental science and pollution research international, 18(9), 1567-1573 (2011-05-18)
This study investigated the effect of copper on chemical oxygen demand (COD) removal efficiency and on the properties (mainly settling and dewatering) and the composition of extracellular polymeric substances (EPS) when 20 mg/L Cu(II) was continuously dosed to a sequencing
Yue Mu et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 83(1), 130-135 (2011-09-06)
The toxicity of sodium benzoate to trypsin was investigated by fluorescence spectroscopy, synchronous fluorescence spectroscopy, UV-visible absorption spectroscopy and circular dichroism (CD) spectroscopy under mimic physiological conditions. Sodium benzoate could unfold trypsin by decreasing the β-sheet structure, which leads to
Sandesh C S Nagamani et al.
Genetics in medicine : official journal of the American College of Medical Genetics, 14(5), 501-507 (2012-01-14)
The urea cycle consists of six consecutive enzymatic reactions that convert waste nitrogen into urea. Deficiencies of any of these enzymes of the cycle result in urea cycle disorders (UCDs), a group of inborn errors of hepatic metabolism that often
Trelita de Sousa et al.
Bioresource technology, 123, 256-262 (2012-09-04)
Pseudomonas nitroreducens TSB.MJ10 exhibiting growth and bioemulsifier production with 0.5% sodium benzoate as the sole carbon source was isolated from a mangrove ecosystem in the vicinity of a petroleum pump. The bioemulsifier is a lipopeptide that is stable over a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service