Skip to Content
Merck
All Photos(1)

Key Documents

28-1770

Sigma-Aldrich

(+)-Sodium L-ascorbate

SAJ special grade, ≥99.0%

Synonym(s):

L(+)-Ascorbic acid sodium salt, Vitamin C sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7NaO6
CAS Number:
Molecular Weight:
198.11
Beilstein:
3767246
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:

grade

SAJ special grade

Assay

≥99.0%

form

solid

availability

available only in Japan

color

white

mp

220 °C (dec.) (lit.)

storage temp.

20-25°C

SMILES string

[Na+].OC[C@H](O)[C@H]1OC(=O)C(O)=C1[O-]

InChI

1S/C6H8O6.Na/c7-1-2(8)5-3(9)4(10)6(11)12-5;/h2,5,7-10H,1H2;/q;+1/p-1/t2-,5+;/m0./s1

InChI key

PPASLZSBLFJQEF-RXSVEWSESA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aleksander Hinek et al.
Journal of dermatological science, 75(3), 173-182 (2014-07-13)
Vitamin C (L-ascorbic acid), a known enhancer of collagen deposition, has also been identified as an inhibitor of elastogenesis. Present studies explored whether and how the L-ascorbic acid derivative (+) sodium L-ascorbate (SA) would affect production of collagen and elastic
Yongliang Zhang et al.
The Journal of biological chemistry, 290(24), 14875-14883 (2015-04-30)
Obesity and metabolic disorders such as insulin resistance and type 2 diabetes have become a major threat to public health globally. The mechanisms that lead to insulin resistance in type 2 diabetes have not been well understood. In this study
Martin Sillesen et al.
The journal of trauma and acute care surgery, 76(1), 12-19 (2013-12-26)
Traumatic brain injury (TBI) and hemorrhagic shock (HS) can be associated with coagulopathy and inflammation, but the mechanisms are poorly understood. We hypothesized that a combination of TBI and HS would disturb coagulation, damage the endothelium, and activate inflammatory and
Lin Zhang et al.
Biosensors & bioelectronics, 69, 77-82 (2015-02-24)
A novel label-free nanosensor has been developed for detecting biothiols including cysteine, glutathione and homocysteine based on poly(thymine)-templated fluorescent copper nanoparticles (CuNPs), which were controlled through thymine-Hg(II)-thymine coordination. This assay provides a simple, cost-effective, and sensitive platform for the detection
Max Leenders et al.
International journal of cancer, 135(12), 2930-2939 (2014-04-29)
Carotenoids and vitamins A, C and E are possibly associated with a reduced colorectal cancer (CRC) risk through antioxidative properties. The association of prediagnostic plasma concentrations and dietary consumption of carotenoids and vitamins A, C and E with the risk

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service