Skip to Content
Merck
All Photos(1)

Key Documents

24-1520

Sigma-Aldrich

Phenol stock solution

100 mg/dL, standard

Synonym(s):

Phenol solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6O
CAS Number:
Molecular Weight:
94.11
Beilstein:
6428463
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
grade:
standard

grade

standard

availability

available only in Japan

concentration

100 mg/dL

dilution

(for analytical testing)

SMILES string

Oc1ccccc1

InChI

1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H

InChI key

ISWSIDIOOBJBQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nina Sanina et al.
FEBS letters, 587(14), 2260-2265 (2013-06-08)
The present work aimed to compare the effects of different lysophosphatidylethanolamine (LPE) content in lipids derived from Yersinia pseudotuberculosis cells exposed and not exposed to phenol on the conformation of OmpF-like porin of these bacteria. Differential scanning calorimetry and intrinsic
Amani M D Elahwany et al.
Acta biologica Hungarica, 64(1), 106-117 (2013-04-10)
Six isolates with phenol degrading ability were obtained from marine sediments by enrichment procedures and an isolate, AM4, was identified as Alcaligenes sp. by 16S rDNA sequencing. The Plackett-Burman design was applied to estimate the significance of culture medium components
B K Shormanov et al.
Sudebno-meditsinskaia ekspertiza, 56(2), 33-37 (2013-06-27)
The present work was designed to study the specific features of 2-methyl hydroxybezene and 3-methyl hydroxybenzene distribution after intragastric administration of these toxicants to warm-blooded animals (rats). They were detected in the unmetabolized form in the internal organs and blood
Fusheng Jiang et al.
PloS one, 8(2), e58004-e58004 (2013-03-08)
This study was carried out to evaluate the utilization probability of the fibrous root part (FRP) of Bletilla striata, which was usually discarded and harvesting pseudobulb part (PSP). The chemical composition, total phenolic content, DPPH radical scavenging activity, Ferric-reducing antioxidant
Hichem Alimi et al.
General physiology and biophysics, 32(1), 115-127 (2013-03-28)
Rhus tripartitum (sumac) is an Anacardiaceae tree with a wide phytotherapeutic application including the use of its roots in the management of gastric ulcer. In the present study the Rhus tripartitum root barks extract (RTE) was phytochemical studied, in vitro

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service