Skip to Content
Merck
All Photos(1)

Key Documents

24-0630

Sigma-Aldrich

Paraformaldehyde

CP

Synonym(s):

Polyoxymethylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2O)nH
CAS Number:
Molecular Weight:
30.03 (as monomer)
MDL number:
UNSPSC Code:
12352104
grade:
CP
technique(s):
cell culture | mammalian: suitable
vapor pressure:
<1.45 mmHg ( 25 °C)

grade

CP

vapor density

1.03 (vs air)

vapor pressure

<1.45 mmHg ( 25 °C)

Assay

≥90% (HCHO)

form

solid

autoignition temp.

572 °F

expl. lim.

73 %

availability

available only in Japan

technique(s)

cell culture | mammalian: suitable

pH

4.0-5.5

mp

120-170 °C (lit.)

density

0.88 g/mL at 25 °C (lit.)

SMILES string

[O-]#[C+H2]

InChI

1S/CH2O/c1-2/h1H2

InChI key

WSFSSNUMVMOOMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Flam. Sol. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ilya Levental et al.
Proceedings of the National Academy of Sciences of the United States of America, 108(28), 11411-11416 (2011-06-29)
Biological membranes are compartmentalized for functional diversity by a variety of specific protein-protein, protein-lipid, and lipid-lipid interactions. A subset of these are the preferential interactions between sterols, sphingolipids, and saturated aliphatic lipid tails responsible for liquid-liquid domain coexistence in eukaryotic
Lili Wang et al.
Cytometry. Part A : the journal of the International Society for Analytical Cytology, 81(7), 567-575 (2012-04-28)
To transform the linear fluorescence intensity scale obtained with fluorescent microspheres to an antibody bound per cell (ABC) scale, a biological cell reference material is needed. Optimally, this material should have a reproducible and tight ABC value for the expression
Geralda A F van Tilborg et al.
Neoplasia (New York, N.Y.), 10(12), 1459-1469 (2008-12-03)
Angiogenic, that is, newly formed, blood vessels play an important role in tumor growth and metastasis and are a potential target for tumor treatment. In previous studies, the alpha(v)beta(3) integrin, which is strongly expressed in angiogenic vessels, has been used
Robyn Mary Brown et al.
PloS one, 5(12), e15889-e15889 (2011-01-07)
Relapse prevention represents the primary therapeutic challenge in the treatment of drug addiction. As with humans, drug-seeking behaviour can be precipitated in laboratory animals by exposure to a small dose of the drug (prime). The aim of this study was
Hans Skvara et al.
The Journal of clinical investigation, 118(9), 3151-3159 (2008-08-09)
PKC isoforms tau, alpha, and beta play fundamental roles in the activation of T cells and other immune cell functions. Here we show that the PKC inhibitor AEB071 both abolishes the production of several cytokines by activated human T cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service