Recommended Products
grade
CP
form
oil
availability
available only in Japan
refractive index
n20/D 1.4795 (lit.)
density
0.93 g/mL at 25 °C (lit.)
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
10 - Combustible liquids
WGK
awg
Flash Point(F)
>230.0 °F
Flash Point(C)
> 110 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Jiqu Xu et al.
Lipids in health and disease, 11, 148-148 (2012-11-02)
Atherosclerosis is a major manifestation of the pathophysiology underlying cardiovascular disease. Flaxseed oil (FO) and α-lipoic acid (LA) have been reported to exert potential benefit to cardiovascular system. This study tried to assess the effect of supplement of FO and
Michael D Breed et al.
Journal of economic entomology, 105(4), 1145-1148 (2012-08-30)
Fatty acids, normally found in comb wax, have a strong influence on nestmate recognition in honey bees, Apis mellifera L. Previous work has shown that bees from different colonies, when treated with 16- or 18-carbon fatty acids, such as oleic
L Salcedo-Sandoval et al.
Meat science, 93(3), 757-766 (2012-12-25)
Nutritional, sensory and technological properties of frankfurters as affected by reformulation processes designed to reduce fat content and improve fatty acid profile were investigated. Healthier oils stabilized in oil in water emulsion or in konjac matrix gel were used as
Joana R N Lemos et al.
Nutrition research (New York, N.Y.), 32(12), 921-927 (2012-12-19)
Malnutrition and chronic inflammation in dialysis patients negatively impact their survival prognosis, and nutrients, such as omega-3 oils, are postulated to reduce proinflammatory response. In this randomized, double-blind, multicenter, placebo-controlled trial, we investigated the effects of flaxseed oil (FO) on
M Ebrahimi et al.
BioMed research international, 2013, 194625-194625 (2013-03-14)
This study was conducted to determine the effects of feeding oil palm frond silage based diets with added linseed oil (LO) containing high α -linolenic acid (C18:3n-3), namely, high LO (HLO), low LO (LLO), and without LO as the control
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service