Skip to Content
Merck
All Photos(1)

Key Documents

19-4530

Sigma-Aldrich

Mineral spirits

CP

Synonym(s):

Mineral turpentine, Ligroin, Petroleum spirits

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12000000
grade:
CP
bp:
179-210 °C (lit.)
vapor pressure:
2 psi ( 21.1 °C)

grade

CP

vapor density

>1 (vs air)

vapor pressure

2 psi ( 21.1 °C)

form

liquid

availability

available only in Japan

refractive index

n20/D 1.424 (lit.)

bp

179-210 °C (lit.)

density

0.752 g/mL at 25 °C (lit.)

SMILES string

C(CCCCCCC)(C)C

InChI

1S/C10H22/c1-4-5-6-7-8-9-10(2)3/h10H,4-9H2,1-3H3

InChI key

SGVYKUFIHHTIFL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

107.6 °F

Flash Point(C)

42 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service